Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MEIPVPVQP SWLRRASAPL LGLSAPGRLF DQRFGEGLLE AELAALCPTT LAPYYLRAPS VALPVAQVPT DPGHFSVLLD VKHFSPEEIA VKVVGEHVEV HARHEERPDE HGFVAREFHR RYRLPPGVDP AAVTSALSPE GVLSIQAAPA SAQAPPPAAA K (1-160 aa encoded by BC068046)
HSPB6 is associated with actin and modulates smooth muscle relaxation. This locus encodes a heat shock protein. The encoded protein likely plays a role in smooth muscle relaxation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: cardiac heat shock protein p20; cardiac p20; epididymis luminal protein 55; Heat shock 20 kDa-like protein p20; heat shock 20-kDa protein; Heat shock protein; Heat shock protein beta-6; heat shock protein family B (small) member B6; Heat shock protein family B member 6; heat shock protein, alpha-crystallin-related, B6; HSP; Hsp20; HspB6; protein phosphatase 1, regulatory subunit 91
Gene Aliases: AA387366; Gm479; HEL55; Hsp20; HSPB6; p20; PPP1R91
UniProt ID: (Human) O14558, (Rat) P97541, (Mouse) Q5EBG6
Entrez Gene ID: (Human) 126393, (Rat) 192245, (Mouse) 243912
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support