Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Immunogen sequence: MEIPVPVQPS WLRRASAPLP GLSAPGRLFD QRFGEGLLEA ELAALCPTTL APYYLRAPSV ALPVAQVPTD PGHFSVLLDV KHFSPEEIAV KVVGEHVEVH ARHEERPDEH GFVAREFHRR YRLPPGVDPA AVTSALSPEG VLSIQAAPAS AQAPPPAAAK; Positive Samples: Mouse lung; Cellular Location: Cytoplasm, Nucleus
HSPB6 is associated with actin and modulates smooth muscle relaxation. This locus encodes a heat shock protein. The encoded protein likely plays a role in smooth muscle relaxation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: cardiac heat shock protein p20; cardiac p20; Heat shock protein; Heat shock protein beta-6; HSP; HspB6
Gene Aliases: AA387366; Gm479; Hsp20; Hspb6
UniProt ID: (Mouse) Q5EBG6
Entrez Gene ID: (Mouse) 243912
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support