Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 4
0.05 µg/mL of MA5-45109 was sufficient for detection of HSP60 in 20 µg of heat shocked HeLa cell lysate by colorimetric immunoblot analysis using goat anti-mouse IgG as the secondary antibody. |Detects approximately 60kDa.
This gene encodes a member of the chaperonin family. The encoded mitochondrial protein may function as a signaling molecule in the innate immune system. This protein is essential for the folding and assembly of newly imported proteins in the mitochondria. This gene is adjacent to a related family member and the region between the 2 genes functions as a bidirectional promoter. Several pseudogenes have been associated with this gene. Two transcript variants encoding the same protein have been identified for this gene. Mutations associated with this gene cause autosomal recessive spastic paraplegia 13.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 60 kDa chaperonin; 60 kDa heat shock protein; 60 kDa heat shock protein, mitochondrial; 60-kDa heat shock protein, mitochondrial; CG12101-PA; CG12101-PB; chaperonin; Chaperonin 60; CPN60; heat shock 60 kDa protein 1; heat shock 60kD protein 1 (chaperonin); heat shock 60kDa protein 1; heat shock 60kDa protein 1 (chaperonin); Heat shock protein; heat shock protein 1 (chaperonin); Heat shock protein 60; heat shock protein 60 (liver); heat shock protein 60 kDa; Heat shock protein 60A; heat shock protein 65; Heat shock protein family D member 1; heat shock protein, 60 kDa; HSP; Hsp 60; HSP-60; HSP-65; hsp60; Hsp60A-PA; Hsp60A-PB; Hsp60s1; HuCHA60; lethal(1)10Ac; mitochondrial heat shock 60 kDa protein 1; Mitochondrial matrix protein P1; P1 protein; P60 lymphocyte protein; short heat shock protein 60 Hsp60s1
Gene Aliases: 12; 60kDa; BP5; CG12101; chaperonin; CPN60; d-hsp60; Dm10A; Dmel\CG12101; Dmel_CG12101; DmHsp60; G62; GROEL; HLD4; Hsp 60; HSP-60; HSP60; HSP60A; Hsp64; HSP65; HSPD1; Hspd1-30p; HuCHA60; I79_011398; IEF16; l(1)10Ac; l(1)BP5; l(1)dp025; l(1)G8; l(1)HM21; l(1)L12; Mmp-P1; RCJMB04_7g5; SPG13; SSP 7506; XELAEV_18045048mg
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support