Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
NeoBiotechnologies
Positive Control: Human spleen stomach or pancreas tissue. MCF-7 cells.
Cellular Location: Cytoplasm. Melanosome.
HSP90 proteins are highly conserved molecular chaperones that have key roles in signal transduction, protein folding, protein degradation, and morphologic evolution. HSP90 proteins normally associate with other cochaperones and play important roles in folding newly synthesized proteins or stabilizing and refolding denatured proteins after stress. There are 2 major cytosolic HSP90 proteins, HSP90AA1, an inducible form, and HSP90AB1 and mitochondria (Chen et al., 2005 ).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: epididymis luminal secretory protein 52; epididymis secretory sperm binding protein Li 65p; Heat shock 84 kDa; Heat shock 86 kDa; heat shock 90kD protein 1, alpha; heat shock 90kD protein 1, alpha-like 4; heat shock 90kD protein 1, beta; heat shock 90kD protein, alpha-like 4; heat shock 90kDa protein 1, alpha; Heat shock protein; heat shock protein 90 kDa; heat shock protein 90kDa alpha (cytosolic), class A member 1; heat shock protein 90kDa alpha (cytosolic), class B member 1; heat shock protein 90kDa alpha family class A member 1; heat shock protein 90kDa alpha family class B member 1; Heat shock protein HSP 90-alpha; Heat shock protein HSP 90-beta; HSP; HSP 84; HSP 86; HSP 90; HSP90-beta; LAP-2; Lipopolysaccharide-associated protein 2; LPS-associated protein 2; Renal carcinoma antigen NY-REN-38
Gene Aliases: D6S182; EL52; HEL-S-65p; HSP84; HSP86; Hsp89; HSP89A; Hsp90; HSP90A; HSP90AA1; HSP90AB1; HSP90B; HSP90N; HSPC1; HSPC2; HSPCA; HSPCAL1; HSPCAL4; HSPCB; HSPN; LAP-2; LAP2
UniProt ID: (Human) P07900, (Human) P08238
Entrez Gene ID: (Human) 3320, (Human) 3326
Molecular Function:
Hsp90 family chaperone
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support