Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: DFYTSITRAR FEELCSDLFR STLEPVEKAL RDAKLDKAQI HDVVLVGGST RIPKVQKLLQ DFFNGKELNK SINPDEAVAY GAAVQAAVLM GDKCEKVQDL LLLDVAPLSL GLETAGGVMT TLIQRNATIP TKQTQTFTTY SDNQPGVFIQ VYEGERAMTK DNNLLGRFEL SGIPPAPRGV PQIEVTFDID ANGILSVTAT DRSTGKANKI TITNDKGRLS KEEVERMVHE AEQYKAEDEA QRDRVAAKNS LEAHVFHVKG SLQEESLRDK IPEEDRRKMQ DKCREVLAWL EHNQLAEKEE YEHQKRELEQ ICRPIFSRLY GGPGVPGGSS CGTQARQGDP STGPIIEEVD
In cooperation with other chaperones, Hsp70s stabilize preexistent proteins against aggregation and mediate the folding of newly translated polypeptides in the cytosol as well as within organelles. These chaperones participate in all these processes through their ability to recognize nonnative conformations of other proteins. They bind extended peptide segments with a net hydrophobic character exposed by polypeptides during translation and membrane translocation, or following stress-induced damage.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Heat shock 70 kDa protein 6; Heat shock 70 kDa protein B'; heat shock 70kD protein 6 (HSP70B'); heat shock 70kDa protein 6 (HSP70B'); Heat shock protein; HSP
Gene Aliases: HSP70B'; HSPA6
UniProt ID: (Human) P17066
Entrez Gene ID: (Human) 3310
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support