Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: KVCVNVHSFK PEELMVKTKD GYVEVSGKHE EKQQEGGIVS KNFTKKIQLP AEVDPVTVFA SLSPEGLLII EAPQVPPYST FGESSFNNEL PQDSQEVTCT
The protein encoded by this gene belongs to the superfamily of small heat-shock proteins containing a conservative alpha-crystallin domain at the C-terminal part of the molecule. The expression of this gene in induced by estrogen in estrogen receptor-positive breast cancer cells, and this protein also functions as a chaperone in association with Bag3, a stimulator of macroautophagy. Thus, this gene appears to be involved in regulation of cell proliferation, apoptosis, and carcinogenesis, and mutations in this gene have been associated with different neuromuscular diseases, including Charcot-Marie-Tooth disease.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Alpha-crystallin C chain; DHMN 2; E2-induced gene 1 protein; heat shock 22kDa protein 8; heat shock 27kDa protein 8; Heat shock protein; Heat shock protein beta-8; HMN 2; HSB8; HSP; HSPB 8; HspB8; OTTHUMP00000239768; Protein kinase H11; Small stress protein-like protein HSP22
Gene Aliases: CMT2L; CRYAC; DHMN2; E2IG1; H11; HMN2; HMN2A; HSP22; HSPB8; PP1629
UniProt ID: (Human) Q9UJY1
Entrez Gene ID: (Human) 26353
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support