Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Western blot analysis using this antibody detected a band around 110 kDa in various lysates.
Immunogen sequence: MSSEADMEC LNQRPPENPD TDKNVQQDNS EAGTQPQVQT DAQQTSQSPP SPELTSEENK IPDADKANEK KVDQPPEAKK PKIKVVNVEL PIEANLVWQL GKDLLNMYIE TEGKMIMQDK LEKERNDAKN AVEEYVYEFR DKLCGPYEKF ICEQDHQNFL RLLTETEDWL YEEGEDQAKQ AYVDKLEELM KIGTPVKVRF QEAEERPKMF EELGQRLQHY AKIAADFRNK DEKYNHIDES EMKKVEKSVN EVMEWMNNVM NAQAKKSLDQ DPVVRAQEIK TKIKELNNTC EPVVTQPKPK IESPKLERTP NGPNIDKKEE DLEDKNNFGA EPPHQNGECY PNEKNSVNMD LD (508-858 aa encoded by BC037553)
HSPH1 prevents the aggregation of denatured proteins in cells under severe stress, on which the ATP levels decrease markedly. This protein inhibits HSPA8/HSC70 ATPase and chaperone activities.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 42 degrees C-HSP; Antigen NY-CO-25; heat shock 105kD alpha; heat shock 105kD beta; heat shock 105kDa/110kDa protein 1; Heat shock 110 kDa protein; Heat shock protein; Heat shock protein 105 kDa; heat shock protein 110; heat shock protein, 105 kDa; Heat shock-related 100 kDa protein E7I; HSP; HSP-E7I; HSP105 42 C-HSP; NY CO 25
Gene Aliases: 105kDa; AI790491; hsp-E7I; HSP105; HSP105A; HSP105B; HSP110; hsp110/105; HSPH1; KIAA0201; NY-CO-25
UniProt ID: (Human) Q92598, (Mouse) Q61699
Entrez Gene ID: (Human) 10808, (Mouse) 15505
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support