Search Thermo Fisher Scientific
MA1-7595 detects HSV from virus samples.
MA1-7595 has been successfully used in ELISA, Western blot and Immunofluorescence procedures.
The MA1-7595 immunogen is HSV (II) infected cells.
Store at 4ºC short term. For extended storage aliquot and store at -80ºC or below. Avoid freeze-thaw cycles.
Members of the herpes virus family (Herpesviridae), Herpes simplex virus types 1 and 2 (HSV-1 and HSV-2), are ubiquitously contagious to humans. Also known as Human herpes virus 1 and 2 (HHV-1 and -2), respectively, HSV-1 strains produce cold sores, while HSV-2 produce most genital herpes. They can be spread when an infected person is producing and shedding the virus. The virion form of herpes viruses consists of a relatively large double-stranded, linear DNA genome encased within the capsid and wrapped in the viral envelope. The different HSV strains carry over 70 open reading frames (ORFs). HSV has been linked to Alzheimer's disease.