Search Thermo Fisher Scientific
The MA1-7268 antibody reacts with Herpes simplex virus 2 gD in viral and infected tissue samples.
MA1-7268 has been successfully used in ELISA and Immunofluorescence applications.
The MA1-7268 antibody was raised against Herpes simplex type 2 gD antigen.
Members of the herpes virus family (Herpesviridae), Herpes simplex virus types 1 and 2 (HSV-1 and HSV-2), are ubiquitously contagious to humans. Also known as Human herpes virus 1 and 2 (HHV-1 and -2), respectively, HSV-1 strains produce cold sores, while HSV-2 produce most genital herpes. They can be spread when an infected person is producing and shedding the virus. The virion form of herpes viruses consists of a relatively large double-stranded, linear DNA genome encased within the capsid and wrapped in the viral envelope. The different HSV strains carry over 70 open reading frames (ORFs). HSV has been linked to Alzheimer's disease.