Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
The protein encoded by this gene functions as a cofactor for the stimulation of transcriptional elongation by HIV-1 Tat, which binds to the HIV-1 promoter through Tat-TAR interaction. This protein may also serve as a dual-function factor to couple transcription and splicing and to facilitate their reciprocal activation. Alternatively spliced transcript variants have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 17S U2 SnRNP complex component HTATSF1; cofactor required for Tat activation of HIV-1 transcription; HIV TAT specific factor 1; HIV Tat-specific factor 1; HIV Tat-specific factor 1 homolog; RP1-196E23.2; Tat-SF1
Gene Aliases: 1600023H17Rik; 2600017A12Rik; 2700077B20Rik; dJ196E23.2; HTATSF1; RGD1563377; TAT-SF1; TATSF1
UniProt ID: (Human) O43719, (Mouse) Q8BGC0
Entrez Gene ID: (Human) 27336, (Rat) 317612, (Mouse) 72459
Molecular Function: RNA splicing factor RNA metabolism protein RNA processing factor nucleic acid metabolism protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support