Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Sequence of this protein is as follows: MLSSVMAPLW ACILVAAGIL ATDTHHPQDS ALYHLSKQLL QKYHKEVRPV YNWTKATTVY LDLFVHAILD VDAENQILKT SVWYQEVWND EFLSWNSSMF DEIREISLPL SAIWAPDIII NEFVDIERYP DLPYVYVNSS GTIENYKPIQ VVSACSLETY AFPFDVQNCS LTFKSILHTV EDVDLAFLRS PEDIQHDKKA FLNDSEWELL SVSSTYSILQ SSAGGFAQIQ FNVVMRRHPL VYVVSLLIPS IFLMLVDLGS FYLPPNCRAR IVFKTSVLVG YTVFRVNMSN QVPRSVGSTP LIGHFFTICM AFLVLSLAKS IVLVKFLHDE QRGGQEQPFL CLRGDTDADR PRVEPRAQRA VVTESSLYGE HLAQPGTLKE VWSQLQSISN YLQTQDQTDQ QEAEWLVLLS RFDRLLFQSY LFMLGIYTIT LCSLWALWGG V
The product of this gene belongs to the ligand-gated ion channel receptor superfamily. This gene encodes subunit B of the type 3 receptor for 5-hydroxytryptamine, a biogenic hormone that functions as a neurotransmitter, a hormone, and a mitogen. This receptor causes fast, depolarizing responses in neurons after activation. It appears that the heteromeric combination of A and B subunits is necessary to provide the full functional features of this receptor, since either subunit alone results in receptors with very low conductance and response amplitude.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 5-HT3-B; 5-hydroxytryptamine (serotonin) receptor 3B, ionotropic; 5-hydroxytryptamine 3 receptor B subunit; 5-hydroxytryptamine receptor 3B; Serotonin receptor 3B; serotonin-gated ion channel subunit
Gene Aliases: 5-HT3B; HTR3B
UniProt ID: (Human) O95264
Entrez Gene ID: (Human) 9177
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support