Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: FQDKQIKDWK KRFIGIRMRT ITPSLVDELK ASNPDFPEVS SGIYVQEVAP NSPSQRGGIQ DGDIIVKVNG RPLVDSSELQ EAVLTESPLL LEVRRGNDDL LFSIAPEVVM
HtrA3 belongs to the human high-temperature requirement factor A (HtrA) family. It is also called pregnancy-related serine protease. HtrA3 has two isoforms (long and short), which is produced by alternative splicing. Similar to HtrA1, HtrA3 binds to various TGF-beta proteins and inhibited the signaling of BMP-4, -2 and TGF-beta 1. HtrA3 does not inhibit signaling originated from a constitutively active BMP receptor, indicating that the inhibition occurred upstream of the cell surface receptor.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: High-temperature requirement factor A3; Pregnancy-related serine protease; probable serine protease HTRA3; Serine protease HTRA3
Gene Aliases: HTRA3; PRSP; Tasp
UniProt ID: (Human) P83110
Entrez Gene ID: (Human) 94031
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support