Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Serine protease.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: High-temperature requirement factor A4; probable serine protease HTRA4; Serine protease HTR4; Serine protease HTRA4
Gene Aliases: B430206E18Rik; HTRA4; RGD1306242
UniProt ID: (Human) P83105, (Rat) D3ZKF5, (Mouse) A2RT60
Entrez Gene ID: (Human) 203100, (Rat) 306564, (Mouse) 330723
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support