Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Immunogen sequence: MATWDEKAV TRRAKVAPAE RMSKFLRHFT VVGDDYHAWN INYKKWENEE EEEEEEQPPP TPVSGEEGRA AAPDVAPAPG PAPRAPLDFR GMLRKLFSSH RFQV (1-103 aa encoded by BC032672)
HVCN1 (hydrogen voltage-gated channel 1), a voltage gated proton channel protein that is highly expressed in immune tissues, mediates the proton conductances required by phagocytic leukocytes for the oxidative burst that underlies microbial killing. HVCN1 moderates the voltage-dependent proton permeability of excitable membranes by allowing the flow of protons in accordance to their electrochemical gradient. HVCN1 is sensitive to zinc ions, and can be inhibited by them. HVCN1 may represent a new therapeutic target for B cell malignancies on continued signaling via the B cell antigen receptor.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: HV1; Hydrogen voltage-gated channel 1; Voltage sensor domain-only protein; Voltage-gated hydrogen channel 1
Gene Aliases: HV1; HVCN1; UNQ578/PRO1140; VSOP
UniProt ID: (Human) Q96D96
Entrez Gene ID: (Human) 84329
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support