Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: GVPQNVSLW AHRKMLQKRV EHYIRTQESA GLAVIDWEDW RPVWVRNWQD KDVYRRLSRQ LVASRHPDWP PDRIVKQAQY EFEFAAQQFM LETLRYVKAV RPRHLWGFYL FPDCYNHDYV QNWESYTGRC PDVEVARNDQ LAWLWAESTA LFPSVYLDET LASSRHGRNF VSFRVQEALR VARTHHANHA LPVYVFTRPT YSRRLTGLSE MDLISTIGES AALGAAGVIL WGDAGYTTST ETCQYLKDYL TRLLVPYVVN VSWATQYCSR AQCHGHGRCV RRNPSASTFL HLSTNSFRLV PGHAPGEPQL RPVGELSWAD IDHLQTHFRC QCYLGWSGEQ CQWDHRQAAG G (99-448 aa encoded by BC000692)
This gene encodes a weak acid-active hyaluronidase. The encoded protein is similar in structure to other more active hyaluronidases. Hyaluronidases degrade hyaluronan, one of the major glycosaminoglycans of the extracellular matrix. Hyaluronan and fragments of hyaluronan are thought to be involved in cell proliferation, migration and differentiation. Although it was previously thought to be a lysosomal hyaluronidase that is active at a pH below 4, the encoded protein is likely a GPI-anchored cell surface protein. This hyaluronidase serves as a receptor for the oncogenic virus Jaagsiekte sheep retrovirus. The gene is one of several related genes in a region of chromosome 3p21.3 associated with tumor suppression. This gene encodes two alternatively spliced transcript variants which differ only in the 5' UTR.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Hyal-2; hyaluronidase 2; Hyaluronidase-2; Hyaluronoglucosaminidase-2; LuCa-2; Lung carcinoma protein 2; lysosomal hyaluronidase; PH-20 homolog; PH20 homolog
Gene Aliases: AI256841; AU020858; AW555733; HYAL2; LUCA2
UniProt ID: (Human) Q12891, (Mouse) O35632
Entrez Gene ID: (Human) 8692, (Rat) 64468, (Mouse) 15587
Molecular Function:
glycosidase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support