Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This polyclonal antibody is raised against 29-460 aa of human HYAL4. The apparent molecular weight is larger than the calculated molecular weight of 54 kDa, which is likely due to posttranslational modification, presumably glycosylation.
Immunogen sequence: LKSISCLKP ARLPIYQRKP FIAAWNAPTD QCLIKYNLRL NLKMFPVIGS PLAKARGQNV TIFYVNRLGY YPWYTSQGVP INGGLPQNIS LQVHLEKADQ DINYYIPAED FSGLAVIDWE YWRPQWARNW NSKDVYRQKS RKLISDMGKN VSATDIEYLA KVTFEESAKA FMKETIKLGI KSRPKGLWGY YLYPDCHNYN VYAPNYSGSC PEDEVLRNNE LSWLWNSSAA LYPSIGVWKS LGDSENILRF SKFRVHESMR ISTMTSHDYA LPVFVYTRLG YRDEPLFFLS KQDLVSTIGE SAALGAAGIV IWGDMNLTSS KANCTKVKQF VSSDLGSYIA NVTRAAEVCS LHLCRNNGRC IRKMWNAPSY LHLNPASYHI EASEDGEFTV KGKASDTDLA VMADTFSCHC YQGYEGADCR EIKTADGCSG VSP (29-460 aa encoded by BC104788)
Endo-hyaluronidase that degrades hyaluronan to smaller oligosaccharide fragments. Has also chondroitin sulfate hydrolase activity, The best substrate being the galactosaminidic linkage in the sequence of a trisulfated tetrasaccharide.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Chondroitin sulfate endo-beta-N-acetylgalactosaminidase; Chondroitin sulfate hydrolase; CSHY; Hyal-4; hyaluronidase 4; Hyaluronidase-4; Hyaluronoglucosaminidase-4
Gene Aliases: 4632428M18Rik; CSHY; HYAL4
UniProt ID: (Human) Q2M3T9, (Mouse) Q05A56
Entrez Gene ID: (Human) 23553, (Rat) 404783, (Mouse) 77042
Molecular Function: glycosidase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support