Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Sandwich ELISA (Recombinant protein).
Immunogen sequence: FVSSDLGSYI ANVTRAAEVC SLHLCRNNGR CIRKMWNAPS YLHLNPASYH IEASEDGEFT VKGKASDTDL AVMADTFSCH CYQGYEGADC REIKTADGCS
Endo-hyaluronidase that degrades hyaluronan to smaller oligosaccharide fragments. Has also chondroitin sulfate hydrolase activity, The best substrate being the galactosaminidic linkage in the sequence of a trisulfated tetrasaccharide.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Chondroitin sulfate endo-beta-N-acetylgalactosaminidase; Chondroitin sulfate hydrolase; CSHY; Hyal-4; hyaluronidase 4; Hyaluronidase-4; Hyaluronoglucosaminidase-4
Gene Aliases: CSHY; HYAL4
UniProt ID: (Human) Q2M3T9
Entrez Gene ID: (Human) 23553
Molecular Function: glycosidase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support