Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: ALGCPRIHLM AGRVPQGADR IAVKAEMEAV FLENLRHAAG VLAQEDLVGL LEPINTRITD PQYFLDTPQQ AAAILQKVGR PNLQLQMDIF HWQIMDGNLT GNIREFLPIV GHVQVAQVPG RGEPSSPGEL NFPYLFQLLE DEGYKGFVGC
Highest antigen sequence identity to the following orthologs: Mouse - 21%, Rat - 91%.
Catalyzes the reversible isomerization between hydroxypyruvate and 2-hydroxy-3-oxopropanoate (also termed tartronate semialdehyde).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Endothelial cell apoptosis protein E-CE1; hydroxypyruvate isomerase homolog; Putative hydroxypyruvate isomerase
Gene Aliases: HT036; HYI; SB156
UniProt ID: (Human) Q5T013
Entrez Gene ID: (Human) 81888
Molecular Function: isomerase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support