Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
FIGURE: 1 / 1
Immunogen sequence: PGRQAAFREG LEQAVRYAKA LGCPRIHLMA GRVPQGADRI AVKAEMEAVF LENLRHAAGV LAQEDLVGLL EPINTRITDP QYFLDTPQQA AAILQKVGRP NLQLQMDIFH WQIMDGNLTG NIREFLPIVG HVQVAQVPGR GEPSSPGELN FPYLFQLLED EGYKGFVGCE YQPRGDTVEG L; Positive Samples: Mouse liver, Mouse lung, Rat spleen
Catalyzes the reversible isomerization between hydroxypyruvate and 2-hydroxy-3-oxopropanoate (also termed tartronate semialdehyde).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Endothelial cell apoptosis protein E-CE1; hydroxypyruvate isomerase; hydroxypyruvate isomerase homolog; Putative hydroxypyruvate isomerase
Gene Aliases: 2700033B16Rik; 6430559E15Rik; HT036; HYI; RGD1561416; SB156
UniProt ID: (Human) Q5T013, (Mouse) Q8R1F5
Entrez Gene ID: (Human) 81888, (Mouse) 68180, (Rat) 500536
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support