Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Positive Samples: HeLa, HepG2, BxPC-3, Mouse testis, Rat testis
Immunogen sequence: MADKVRRQRP RRRVCWALVA VLLADLLALS DTLAVMSVDL GSESMKVAIV KPGVPMEIVL NKESRRKTPV IVTLKENERF FGDSAASMAI KNPKATLRYF QHLLGKQADN PHVALYQARF PEHELTFDPQ RQTVHFQISS QLQFSPEEVL
HYOU1 belongs to the heat shock protein 70 family. The protein is thought to play an important role in protein folding and secretion in the ER. Since suppression of the protein is associated with accelerated apoptosis, it is also suggested to have an important cytoprotective role in hypoxia-induced cellular perturbation. This protein has been shown to be up-regulated in tumors, especially in breast tumors, and thus it is associated with tumor invasiveness. This signal peptide-lacking protein, which is only 3 amino acids shorter than the mature protein in the ER, is thought to have a housekeeping function in the cytosol. In rat, this protein localizes to both the ER by a carboxy-terminal peptide sequence and to mitochondria by an amino-terminal targeting signal.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 140 kDa Ca(2+)-binding protein; 150 kDa oxygen-regulated protein; 150-kDa oxygen regulated protein; 170 kDa glucose regulated protein GRP170; 170 kDa glucose-regulated protein; calcium binding protein 140; calcium binding protein, 140 kDa; CBP-140; DKFZp686N08236; FLJ94899; FLJ97572; Grp 170; GRP-170; HSP 12A; Hypoxia up-regulated protein 1; Orp 150; ORP-150; oxygen regulated protein (150kD)
Gene Aliases: AI415631; Cab140; CBP-140; GRP-170; GRP170; HSP12A; HYOU1; ORP-150; ORP150
UniProt ID: (Human) Q9Y4L1, (Rat) Q63617, (Mouse) Q9JKR6
Entrez Gene ID: (Human) 10525, (Rat) 192235, (Mouse) 12282
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support