- Order Status
- Quick Order
-
Don't have an account ? Create Account
The recommended shelf life for this product is 1 year from date of receipt.
Based on 100% sequence identity, this antibody is predicted to react with Gorilla
GSG2 (haspin) is a conserved kinase required for mitosis. GSG2 phosphorylates histone H3 and is viewed as a potential therapeutic target for oncology. GSG2 is required for normal alignment of chromosomes at metaphase.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: germ cell associated 2 (haspin); Germ cell-specific gene 2 protein; H-haspin; Haploid germ cell-specific nuclear protein kinase; Serine/threonine-protein kinase haspin
Gene Aliases: GSG2; HASPIN
UniProt ID: (Human) Q8TF76
Entrez Gene ID: (Human) 83903
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support