Search Thermo Fisher Scientific
HEV ORF3 is a transmembrane protein localized at ER membranes (20) and forms multimeric complexes, presumably through homophilic interactions. ORF3 is a predicted transmembrane protein that associates with ER membranes and can form homooligomers.