Search Thermo Fisher Scientific
Rockland
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Store vial at -20° C prior to opening. Aliquot contents and freeze at -20° C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4° C as an undiluted liquid. Dilute only prior to immediate use.
Directed against the MHV-A59 nsp9 protein. No cross reactivity occurs with SARS CoV nsp9. Cross reactivity with homologues from other sources has not been tested.
The nonstructural protein 9 (nsp9) is one of the Mouse hepatitis virus replicase cleavage products, encoded by ORF1a. Nsp9 is an RNA-binding protein that is thought to be part of the viral replication complex, which is associated with intracellular membranes.