Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Hepsin is a cell surface serine protease.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: hepsin (transmembrane protease, serine 1); Serine protease hepsin; testicular tissue protein Li 85; Transmembrane protease serine 1
Gene Aliases: Hlb320; HPN; TMPRSS1
UniProt ID: (Human) P05981, (Rat) Q05511, (Mouse) O35453
Entrez Gene ID: (Human) 3249, (Rat) 29135, (Mouse) 15451
Molecular Function:
serine protease
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support