Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Highest antigen sequence indentity to the following orthologs: Mouse (67%), Rat (67%).
This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-60222. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
This gene encodes a member of the beta subfamily of defensins. Beta-defensins are antimicrobial peptides that protect tissues and organs from infection by a variety of microorganisms. This gene is found in a cluster with other beta-defensin genes on the long arm of chromosome 20. Multiple transcript variants encoding different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Beta-defensin 119; Beta-defensin 120; Beta-defensin 19; Beta-defensin 20; DEFB-19; DEFB-20; Defensin, beta 119; Defensin, beta 120; defensin, beta 19; defensin, beta 20; ESC42-RELA
Gene Aliases: DEFB-19; DEFB-20; DEFB119; DEFB120; DEFB19; DEFB20; ESC42-RELA; ESC42-RELB; UNQ2449/PRO5729
UniProt ID: (Human) Q8N690
Entrez Gene ID: (Human) 245932
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support