Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Highest antigen sequence indentity to the following orthologs: Mouse (90%), Rat (90%).
This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-85031. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
DnaJ is a testis-specific heat shock protein and functions in association with DnaK(Hsp70). It is a molecular chaperone to facilitate protein folding. DnaJ plays a key role in the chaperone reaction by stimulating the ATPase activity and activating the substrate binding of Hsp70.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: beta cysteine string protein; beta-CSP; CSP-beta; cysteine string protein beta; Cysteine-string protein isoform beta; DnaJ (Hsp40) homolog, subfamily C, member 5 beta; DnaJ homolog subfamily C member 5B; dnaJ homolog subfamily C member X; testicular tissue protein Li 55
Gene Aliases: CSP-beta; CSPBETA; DNAJC5B
UniProt ID: (Human) Q9UF47
Entrez Gene ID: (Human) 85479
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support