Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Highest antigen sequence indentity to the following orthologs: Mouse (76%), Rat (76%).
This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-63792. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Elastin microfibril interface-located protein 3; Elastin microfibril interface-located protein 5; Elastin microfibril interfacer 3; Elastin microfibril interfacer 5; EMILIN-3; EMILIN-5
Gene Aliases: C20orf130; dJ620E11.4; EMILIN3; EMILIN5
UniProt ID: (Human) Q9NT22
Entrez Gene ID: (Human) 90187
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support