Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Highest antigen sequence indentity to the following orthologs: Mouse (74%), Rat (74%).
This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-67448. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
Iron-regulatory hormone that acts as an erythroid regulator after hemorrhage: produced by erythroblasts following blood loss and mediates suppression of hepcidin (HAMP) expression in the liver, thereby promoting increased iron absorption and mobilization from stores. Promotes lipid uptake into adipocytes and hepatocytes via transcriptional up-regulation of genes involved in fatty acid uptake.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Complement C1q tumor necrosis factor-related protein 15; Erythroferrone; family with sequence similarity 132 member B; family with sequence similarity 132, member B; Myonectin; protein FAM132B
Gene Aliases: C1QTNF15; CTRP15; ERFE; FAM132B
UniProt ID: (Human) Q4G0M1
Entrez Gene ID: (Human) 151176
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support