Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
FIGURE: 1 / 3
Positive Samples: Human plasma
Immunogen sequence: APTKAPDVFP IISGCRHPKD NSPVVLACLI TGYHPTSVTV TWYMGTQSQP QRTFPEIQRR DSYYMTSSQL STPLQQWRQG EYKCVVQHTA SKSKKEIFRW
This type I transmembrane protein is co-expressed on the surface of mature naive B cells with membrane IgM. IgD also exists as a soluble form in blood serum. Studies have demonstrated that IgD associates with the B cell receptor and participates in mediating signal transduction upon antigen binding.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Entrez Gene ID: (Human) 3495
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support