Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Highest antigen sequence indentity to the following orthologs: Mouse (73%), Rat (73%).
This recombinant protein control fragment may be used for blocking experiments with the corresponding antibodies, PA5-139736, PA5-52707. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Protein Shroom4; Second homolog of apical protein
Gene Aliases: KIAA1202; SHAP; SHROOM4
UniProt ID: (Human) Q9ULL8
Entrez Gene ID: (Human) 57477
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support