Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Highest antigen sequence indentity to the following orthologs: Mouse (92%), Rat (92%).
This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-63157. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: dJ310O13.1; Probable tubulin polyglutamylase TTLL9; tubulin tyrosine ligase-like family, member 9; Tubulin--tyrosine ligase-like protein 9
Gene Aliases: C20orf125; TTLL9
UniProt ID: (Human) A6NH06
Entrez Gene ID: (Human) 164395
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support