Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
MA1-70066 detects I-Ek in mouse samples.
MA1-70066 has been successfully used in flow cytometry and cytotoxicity studies.
The MA1-70066 immunogen is C3H.
MHC (major histocompatibility complex) class II molecules are transmembrane glycoproteins expressed on the surface of professional antigen-presenting cells, such as macrophages, dendritic cells and B cells. Before their exposition on the cell surface, the MHC class II molecules react with endocytosed exogenous antigens, which are then presented to the T cells. The antigen-binding grove between MHC class II alpha and beta chain is open at both ends and is 15-24 amino acid residues long.
HLA and MHC antibodies play a significant role in Immunopeptidomics, facilitating the identification and characterization of neoantigens through high-performance liquid chromatography coupled to tandem Mass Spectrometry.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: cell surface glycoprotein; H-2 class II histocompatibility antigen, E-D alpha chain; H-2 class II histocompatibility antigen, E-K alpha chain; H-2 class II histocompatibility antigen, E-U alpha chain; H-2 class II histocompatibility antigen, I-E beta chain; H2-IE-alpha; histocompatibility 2, class II antigen E alpha; I-E alpha MHC class II; I-E beta MHC class II; major histocompatibility complex H-2E beta chain; MGC117888; MHC class II antigen E alpha; MHC class II IE-b; MHC class II protein; MHC H2-IE-beta cell surface glycoprotein
Gene Aliases: AI323765; E-alpha-f; H-2Ea; H-2Eb; H2-Ea; H2-Ea-ps; H2Eb; I-Ealpha; Ia-3; Ia-4; Ia3; Ia4
UniProt ID: (Mouse) P04224
Entrez Gene ID: (Mouse) 100504404, (Mouse) 14969
Molecular Function:
major histocompatibility complex protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support