Search Thermo Fisher Scientific
FIGURE: 1 / 2
This product can be used in direct ELISA.
AIF1 is induced by cytokines and interferon. It is thought to be involved in the negative regulation of growth of vascular smooth muscle cells, which contributes to the anti-inflammatory response to vessel wall trauma.