Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Primary Antibodies
  • Search All
  • Popular Product Areas
  • TaqMan Assays
  • Primary Antibodies
  • Secondary Antibodies
  • Isotype Controls
  • Proteins & Peptides
  • ELISA Kits
  • All Documents & Support
  • Certificates
  • SDS
  • Manuals & Protocols
  • Product FAQs
    Search button Close
            Target of Interest
            Application
            Target Species
            • Order Status
            • Quick Order
            • Sign in
              Sign in
              Don't have an account ? Create Account
              • Account
              • Check Order Status
              • Aspire Member Program
              • Connect: Lab, Data, Apps
              • Custom Products & Projects
              • Services Central

            Disclaimer

            Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

            Loading ...
            • Primary Antibodies ›
            • IBA1 Antibodies

            Invitrogen

            IBA1 Polyclonal Antibody

            View all (54) IBA1 antibodies

            Promotions

            View available promotion(s)

            Promo Code: P5906365 Don't miss out: buy 3, only pay for 2 Extended to June 27: take advantage of our 3 for 2 promotion. Whether you are restocking essentials or trying something new, now is a great time to get more for your money!. Learn more

            Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more

            Datasheet
            Protocols
            Questions & Answers
            Catalog # PA5-143610
            Web quote
            You can generate a web quote (PDF) by signing in to your account and adding products to your cart. View cart and click the 'Generate a web quote' button. With your web quote document, you can get approval for your order and complete your order online.
             
            Price (USD)
            494.00
             
            Estimated availability date: 07-Jul-2025
            Check your price
            Request bulk or custom format
            Join Aspire™ for free antibodies
            Datasheet
            Protocols
            Questions & Answers

            Cite IBA1 Polyclonal Antibody

            • Antibody Testing Data (3)
            Application
            IBA1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            Group 53 Created with Sketch.

            FIGURE: 1 / 3

            IBA1 Antibody (PA5-143610) in IHC (P)

            Immunohistochemistry analysis of IBA1 in rat cerebellar molecular layer (top) and granular layer (bottom). Samples were stained with IBA1 polyclonal antibody (Product # PA5-143610) using a dilution of 1:1,000 (green). Microglia are very small cells with fine processes spreading in three dimensions and so are best visualized in a confocal Z stack. Red shows the processes of Purkinje cells and the perikarya o... View More
            View Product

            IBA1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            IBA1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            IBA1 Antibody in Western Blot (WB)

            Product Details

            PA5-143610

            Applications
            Tested Dilution
            Publications

            Western Blot (WB)

            1:1,000-1:5,000
            -

            Immunohistochemistry (Paraffin) (IHC (P))

            1:1,000-1:2,000
            -

            Immunocytochemistry (ICC/IF)

            1:1,000-1:2,000
            -
            Product Specifications

            Species Reactivity

            Human, Mouse, Rat

            Host/Isotype

            Rabbit / IgG

            Class

            Polyclonal

            Type

            Antibody

            Immunogen

            Peptide identical to the C-terminal of human IBA1 coupled to KLH
            View immunogen

            Conjugate

            Unconjugated

            Form

            Liquid

            Concentration

            Conc. Not Determined

            Storage buffer

            whole serum

            Contains

            5mM sodium azide

            Storage conditions

            Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

            Shipping conditions

            Ambient (domestic); Wet ice (international)

            RRID

            AB_2942839

            Target Information

            Ionized calcium-binding adapter molecule 1 (IBA1), also known by its gene name AIF1, is a protein expressed predominantly by microglia in the brain and spinal cord. This protein belongs to the EF-hand calcium-binding protein family and plays a crucial role in microglial activation and migration in response to brain injury or neuroinflammation. IBA1's function is integral to microglial motility and phagocytic activity, facilitating the cellular response to pathogenic stimuli and promoting tissue homeostasis and repair in the central nervous system. IBA1 serves as a reliable marker for activated microglia in various neurological disorders, including Alzheimer's disease, Parkinson's disease, and multiple sclerosis, where increased expression correlates with disease progression and severity. The protein's structural features enable it to bind calcium ions, inducing conformational changes that activate signaling pathways essential for microglial function. Its expression is highly regulated by inflammatory cytokines, underpinning its role in neuroimmune responses. Due to its specific expression in microglia during pathological conditions, IBA1 is widely used in research as a marker to study microglial status and activity, and it remains a focal point for understanding microglial involvement in neurodegenerative diseases.

            For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

            References (0)

            Have you cited this product in a publication?
            Let us know so we can reference it here.
            Cite this product

            Bioinformatics

            Protein Aliases: AIF-1; Allograft inflammatory factor 1; balloon angioplasty responsive transcript; balloon angioplasty-responsive transcript 1; DADB-70P7.8; DASS-82G15.2; Em:AF129756.17; Iba; interferon gamma responsive transcript; ionized calcium binding adapter molecule 1; Ionized calcium-binding adapter molecule 1; Microglia response factor; MRF-1; protein BART-1; Protein G1; testis specific

            View more View less

            Gene Aliases: AI607846; AIF-1; AIF1; BART-1; Bart1; D17H6S50E; G1; IBA1; IRT-1; IRT1; mrf-1; Mrf1

            View more View less

            UniProt ID: (Human) P55008, (Mouse) O70200, (Rat) P55009

            View more View less

            Entrez Gene ID: (Human) 199, (Mouse) 11629, (Rat) 29427

            View more View less

            Function(s)

            molecular_function calcium ion binding actin filament binding actin binding protein binding metal ion binding calmodulin-related

            Process(es)

            microglial cell activation positive regulation of protein phosphorylation phagocytosis, engulfment inflammatory response negative regulation of gene expression positive regulation of muscle hyperplasia Rac protein signal transduction actin filament polymerization cellular response to extracellular stimulus cellular response to hormone stimulus positive regulation of T cell proliferation negative regulation of apoptotic process positive regulation of nitric oxide biosynthetic process positive regulation of smooth muscle cell proliferation negative regulation of smooth muscle cell proliferation response to axon injury actin filament bundle assembly response to glucocorticoid response to electrical stimulus cellular response to morphine cellular response to interferon-gamma cellular response to hydroperoxide negative regulation of smooth muscle cell chemotaxis positive regulation of smooth muscle cell chemotaxis positive regulation of monocyte chemotaxis ruffle assembly positive regulation of G1/S transition of mitotic cell cycle positive regulation of T cell migration positive regulation of cell migration response to cytokine macrophage activation
            It has to be done as per old AB suggested Products section.
            guarantee_icon

            Performance Guarantee

            If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

            Learn more
            help_icon

            We're here to help

            Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

            Contact tech support
            Ordering Plus Icon Minus Icon
            • Order Status
            • Order Help
            • Quick Order
            • Supply Center
            • eProcurement
            Support Plus Icon Minus Icon
            • Help and Support
            • Contact Us
            • Technical Support Centers
            • Documents and Certificates
            • Report a Site Issue
            Resources Plus Icon Minus Icon
            • Learning Centers
            • Promotions
            • Events and Webinars
            • Social Media
            About Thermo Fisher Plus Icon Minus Icon
            • About Us
            • Careers
            • Investors
            • News
            • Social Responsibility
            • Trademarks
            • Consumer Health Data Privacy Policy
            Our Portfolio Plus Icon Minus Icon
            • Thermo Scientific
            • Applied Biosystems
            • Invitrogen
            • Gibco
            • Ion Torrent
            • Fisher Scientific
            • Unity Lab Services
            • Patheon
            • PPD
            • Terms & Conditions
            • Privacy Information Center
            • Price & Freight Policy
            • Cookie Preferences

              Your choices regarding cookies on this site

              We and our affiliates and vendors use cookies and similar technologies to operate our sites, recognize visitors to our sites, provide secure log-in, collect statistics to optimize site functionality, and deliver content tailored to your interests. Click Accept All to accept all cookies and go directly to the site, click Reject All to reject all but cookies strictly necessary to the functioning of this site (required cookies), or click on Manage Settings to see detailed descriptions of the types of cookies and choose whether to accept certain cookies while on the site.
            © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
            united states flag icon
            United States

            Your items have has been added!


            Host server : magellan-search-green-56756dfb8b-6r49h:80/100.66.79.199:80.
            git-commit: de6905f249d25e22729e613aef254bb544bd8b7c
            git-url: https://github.com/thermofisher/magellan-search
            git-branch: release/2.31.1-2025.06.25-1.0