Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
This Antibody was verified by Knockdown to ensure that the antibody binds to the antigen stated.
FIGURE: 1 / 4
Positive Control: HeLa, A431, mouse brain, rat brain
Predicted Reactivity: Zebrafish (90%), Dog (99%), Rhesus Monkey (100%), Bovine (98%)
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene may belong to a protease family responsible for intercellular peptide signalling. Though its role in the cellular processing of insulin has not yet been defined, insulin-degrading enzyme is thought to be involved in the termination of the insulin response.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Abeta-degrading protease; FLJ35968; Insulin degrading enzyme; Insulin protease; Insulin-degrading enzyme; Insulinase; Insulysin
Gene Aliases: 1300012G03Rik; 4833415K22Rik; AA675336; AI507533; IDE; INSDEGM; INSULYSIN
UniProt ID: (Human) P14735, (Rat) P35559
Entrez Gene ID: (Human) 3416, (Rat) 25700, (Mouse) 15925
Molecular Function:
metalloprotease
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support