Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MSDINLDWVD RRQLQRLEEM LIVVDENDKV IGADTKRNCH LNENIEKGLL HRAFSVVLFN TKNRILIQQR SDTKVTFPGY FTDSCSSHPL YNPAELEEKD AIGVRRAAQR RLQAELGIPG EQISPEDIVF MTIYHHKAKS DRIWGEHEIC YLLLVRKNVT LNPDPSETKS ILYLSQEELW ELLEREARGE VKVTPWLRTI AERFLYRWWP HLDDVTPFVE LHKIHRV
Catalyzes the 1,3-allylic rearrangement of the homoallylic substrate isopentenyl (IPP) to its highly electrophilic allylic isomer, dimethylallyl diphosphate (DMAPP).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: diphosphate dimethylallyl diphosphate isomerase 2; IPP isomerase 2; Isopentenyl pyrophosphate isomerase 2; Isopentenyl-diphosphate delta-isomerase 2
Gene Aliases: IDI2; IPPI2
UniProt ID: (Human) Q9BXS1
Entrez Gene ID: (Human) 91734
Molecular Function: isomerase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support