Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MLHFHYYDT SNKIMEPHRP NVKTAVPLSL ESYHISEEYG FLLPDSLKEL PDHYRPWMEI ANKLPQLIDA HQLQAHVDKM PLLSCQFLKG HREQRLAHLV LSFLTMGYVW QEGEAQPAEV LPRNLALPFV EVSRNL (1-135 aa encoded by B C113496)
Along with the enzymes encoded by the INDO and TDO2 genes, the enzyme encoded by the INDOL1 gene metabolizes tryptophan in the kynurenine pathway.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 3-dioxygenase 2; 3-dioxygenase-like protein 1; EC 1.13.11.; IDO-2; INDOL 1; indoleamine 2; Indoleamine 2,3 dioxygenase 2; Indoleamine 2,3 dioxygenase like 1 protein; Indoleamine 2,3-dioxygenase 2; indoleamine 2,3-dioxygenase-like 1 protein; Indoleamine 2,3-dioxygenase-like protein 1; Indoleamine pyrrole 2,3 dioxygenase like 1; Indoleamine-pyrrole 2; indoleamine-pyrrole 2,3 dioxygenase-like 1; Indoleamine-pyrrole 2,3-dioxygenase-like protein 1
Gene Aliases: IDO2; INDOL1
UniProt ID: (Human) Q6ZQW0
Entrez Gene ID: (Human) 169355
Molecular Function: oxygenase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support