Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 5
Positive Samples: Rat thymus; Cellular Location: Cytoplasm
Immunogen sequence: LLQALRDYKP YGDLVQQ
IFI-44 (interferon-induced protein 44), also known as p44 or MTAP44 (microtubule-associated protein 44), is a 444 amino acid protein that localizes to the cytoplasm and, upon induction by IFN-βs, aggregates to form microtubular structures. Human IFI-44 shares 97% sequence similarity with its chimp counterpart, suggesting a conserved role between species. The gene encoding IFI-44 maps to human chromosome 1, which spans 260 million base pairs, contains over 3,000 genes and comprises nearly 8% of the human genome. Chromosome 1 houses a large number of disease-associated genes, including those that are involved in familial adenomatous polyposis, Stickler syndrome, Parkinson's disease, Gaucher disease, schizophrenia and Usher syndrome.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Interferon-induced protein 44; interferon-induced, hepatitis C-associated microtubular aggregate protein (44kD); Microtubule-associated protein 44; p44; TBC/LysM-associated domain containing 5
Gene Aliases: A430056A10Rik; AW261460; IFI44; MTAP44; p44; TLDC5
UniProt ID: (Human) Q8TCB0, (Mouse) Q8BV66
Entrez Gene ID: (Human) 10561, (Mouse) 99899, (Rat) 310969
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support