Product References
Distinct SARS-CoV-2 specific NLRP3 and IL-1β responses in T cells of aging patients during acute COVID-19 infection.
Frontiers in immunology
Mahalingam SS,Jayaraman S,Arunkumar A,Dudley HM,Anthony DD,Shive CL,Jacobson JM,Pandiyan P
Published figure using IFN gamma monoclonal antibody (Product # 14-7319-81) in Flow Cytometry
Thu Oct 12 00:00:00 EDT 2023
CD8+ T cell-Dependent Remodeling of the Tumor Microenvironment Overcomes Chemoresistance.
Cancer immunology research
Lao L,Zeng W,Huang P,Chen H,Jia Z,Wang P,Huang D,Chen J,Nie Y,Yang L,Wu W,Liu J
Published figure using IFN gamma monoclonal antibody (Product # 14-7319-81) in Flow Cytometry
Wed Mar 01 00:00:00 EST 2023
Rapamycin improves Graves' orbitopathy by suppressing CD4+ cytotoxic T lymphocytes.
JCI insight
Zhang M,Chong KK,Chen ZY,Guo H,Liu YF,Kang YY,Li YJ,Shi TT,Lai KK,He MQ,Ye K,Kahaly GJ,Shi BY,Wang Y
Published figure using IFN gamma monoclonal antibody (Product # 14-7319-81) in Flow Cytometry
Wed Feb 08 00:00:00 EST 2023
HERC2 promotes inflammation-driven cancer stemness and immune evasion in hepatocellular carcinoma by activating STAT3 pathway.
Journal of experimental & clinical cancer research : CR
Liu Y,Xu Q,Deng F,Zheng Z,Luo J,Wang P,Zhou J,Lu X,Zhang L,Chen Z,Zhang Q,Chen Q,Zuo D
Published figure using IFN gamma monoclonal antibody (Product # 14-7319-81) in Flow Cytometry
Wed Feb 01 00:00:00 EST 2023
GPR116 receptor regulates the antitumor function of NK cells via Gαq/HIF1α/NF-κB signaling pathway as a potential immune checkpoint.
Cell & bioscience
Guo D,Jin C,Gao Y,Lin H,Zhang L,Zhou Y,Yao J,Duan Y,Ren Y,Hui X,Ge Y,Yang R,Jiang W
Published figure using IFN gamma monoclonal antibody (Product # 14-7319-81) in Flow Cytometry
Thu Mar 09 00:00:00 EST 2023
GM-CSF-activated human dendritic cells promote type 1 T follicular helper cell polarization in a CD40-dependent manner.
Journal of cell science
Korniotis S,Saichi M,Trichot C,Hoffmann C,Amblard E,Viguier A,Grondin S,Noel F,Mattoo H,Soumelis V
Published figure using IFN gamma monoclonal antibody (Product # 14-7319-81) in Flow Cytometry
Tue Nov 01 00:00:00 EDT 2022
Preventing Surgery-Induced NK Cell Dysfunction Using Anti-TGF-β Immunotherapeutics.
International journal of molecular sciences
Market M,Tennakoon G,Scaffidi M,Cook DP,Angka L,Ng J,Tanese de Souza C,Kennedy MA,Vanderhyden BC,Auer RC
Published figure using IFN gamma monoclonal antibody (Product # 14-7319-81) in Flow Cytometry
Wed Nov 23 00:00:00 EST 2022
LINC01123 promotes immune escape by sponging miR-214-3p to regulate B7-H3 in head and neck squamous-cell carcinoma.
Cell death & disease
Li H,Yang Z,Yang X,Zhang F,Wang J,Wu Z,Wanyan C,Meng Q,Gao W,Yang X,Wei J
Published figure using IFN gamma monoclonal antibody (Product # 14-7319-81) in Flow Cytometry
Thu Feb 03 00:00:00 EST 2022
An Fc-Competent Anti-Human TIGIT Blocking Antibody Ociperlimab (BGB-A1217) Elicits Strong Immune Responses and Potent Anti-Tumor Efficacy in Pre-Clinical Models.
Frontiers in immunology
Chen X,Xue L,Ding X,Zhang J,Jiang L,Liu S,Hou H,Jiang B,Cheng L,Zhu Q,Zhang L,Zhou X,Ma J,Liu Q,Li Y,Ren Z,Jiang B,Song X,Song J,Jin W,Wei M,Shen Z,Liu X,Wang L,Li K,Zhang T
Published figure using IFN gamma monoclonal antibody (Product # 14-7319-81) in Flow Cytometry
Mon May 02 00:00:00 EDT 2022
NRF2/Itaconate Axis Regulates Metabolism and Inflammatory Properties of T Cells in Children with JIA.
Antioxidants (Basel, Switzerland)
Rajendiran A,Subramanyam SH,Klemm P,Jankowski V,van Loosdregt J,Vastert B,Vollbach K,Wagner N,Tenbrock K,Ohl K
Published figure using IFN gamma monoclonal antibody (Product # 14-7319-81) in Flow Cytometry
Thu Dec 08 00:00:00 EST 2022
Therapeutic inhibition of the SRC-kinase HCK facilitates T cell tumor infiltration and improves response to immunotherapy.
Science advances
Poh AR,Love CG,Chisanga D,Steer JH,Baloyan D,Chopin M,Nutt S,Rautela J,Huntington ND,Etemadi N,O'Brien M,O'Keefe R,Ellies LG,Macri C,Mintern JD,Whitehead L,Gangadhara G,Boon L,Chand AL,Lowell CA,Shi W,Pixley FJ,Ernst M
14-7319-81 was used in Flow cytometry/Cell sorting to suggest that therapeutic targeting of HCK activity enhances response to immunotherapy by simultaneously stimulating immune cell activation and inhibiting the immunosuppressive tumor microenvironment.
Fri Jun 24 00:00:00 EDT 2022
A vaccine targeting mutant IDH1 in newly diagnosed glioma.
Nature
Platten M,Bunse L,Wick A,Bunse T,Le Cornet L,Harting I,Sahm F,Sanghvi K,Tan CL,Poschke I,Green E,Justesen S,Behrens GA,Breckwoldt MO,Freitag A,Rother LM,Schmitt A,Schnell O,Hense J,Misch M,Krex D,Stevanovic S,Tabatabai G,Steinbach JP,Bendszus M,von Deimling A,Schmitt M,Wick W
Published figure using IFN gamma monoclonal antibody (Product # 14-7319-81) in Flow Cytometry
Thu Apr 01 00:00:00 EDT 2021
Cancer-secreted exosomal miR-1468-5p promotes tumor immune escape via the immunosuppressive reprogramming of lymphatic vessels.
Molecular therapy : the journal of the American Society of Gene Therapy
Zhou C,Wei W,Ma J,Yang Y,Liang L,Zhang Y,Wang Z,Chen X,Huang L,Wang W,Wu S
Published figure using IFN gamma monoclonal antibody (Product # 14-7319-81) in Flow Cytometry
Wed Apr 07 00:00:00 EDT 2021
Ageing promotes early T follicular helper cell differentiation by modulating expression of RBPJ.
Aging cell
Webb LMC,Fra-Bido S,Innocentin S,Matheson LS,Attaf N,Bignon A,Novarino J,Fazilleau N,Linterman MA
Published figure using IFN gamma monoclonal antibody (Product # 14-7319-81) in Flow Cytometry
Fri Jan 01 00:00:00 EST 2021
Suppression of 4.1R enhances the potency of NKG2D-CAR T cells against pancreatic carcinoma via activating ERK signaling pathway.
Oncogenesis
Gao Y,Lin H,Guo D,Cheng S,Zhou Y,Zhang L,Yao J,Farooq MA,Ajmal I,Duan Y,He C,Tao L,Wu S,Liu M,Jiang W
Published figure using IFN gamma monoclonal antibody (Product # 14-7319-81) in Flow Cytometry
Tue Sep 21 00:00:00 EDT 2021
Dysregulated Peripheral Invariant Natural Killer T Cells in Plaque Psoriasis Patients.
Frontiers in cell and developmental biology
Hu Y,Chen Y,Chen Z,Zhang X,Guo C,Yu Z,Xu P,Sun L,Zhou X,Gong Y,Yu Q,Shi Y
Published figure using IFN gamma monoclonal antibody (Product # 14-7319-81) in Flow Cytometry
Wed Feb 23 00:00:00 EST 2022
Identification of CTL Epitopes on Efflux Pumps of the ATP-Binding Cassette and the Major Facilitator Superfamily of Mycobacterium tuberculosis.
Journal of immunology research
Lin Y,Dong Y,Gao Y,Shi R,Li Y,Zhou X,Liu W,Li G,Qi Y,Wu Y
Published figure using IFN gamma monoclonal antibody (Product # 14-7319-81) in Flow Cytometry
Mon Sep 20 00:00:00 EDT 2021
Early loss of T lymphocyte 4-1BB receptor expression is associated with higher short-term mortality in alcoholic hepatitis.
PloS one
Eriksen LL,Nielsen MA,Laursen TL,Deleuran B,Vilstrup H,Støy S
Published figure using IFN gamma monoclonal antibody (Product # 14-7319-81) in Flow Cytometry
Mon Nov 29 00:00:00 EST 2021
TIGIT+ TIM-3+ NK cells are correlated with NK cell exhaustion and disease progression in patients with hepatitis B virus‑related hepatocellular carcinoma.
Oncoimmunology
Yu L,Liu X,Wang X,Yan F,Wang P,Jiang Y,Du J,Yang Z
Published figure using IFN gamma monoclonal antibody (Product # 14-7319-81) in Flow Cytometry
Mon Aug 02 00:00:00 EDT 2021
A protocol for isolation of primary human immune cells from the liver and mesenteric white adipose tissue biopsies.
STAR protocols
Moreno-Fernandez ME,Damen MSMA,Divanovic S
Published figure using IFN gamma monoclonal antibody (Product # 14-7319-81) in Flow Cytometry
Fri Dec 17 00:00:00 EST 2021
Suppression of mucosal Th17 memory responses by acellular pertussis vaccines enhances nasal Bordetella pertussis carriage.
NPJ vaccines
Dubois V,Chatagnon J,Thiriard A,Bauderlique-Le Roy H,Debrie AS,Coutte L,Locht C
Published figure using IFN gamma monoclonal antibody (Product # 14-7319-81) in Flow Cytometry
Fri Jan 08 00:00:00 EST 2021
Niraparib exhibits a synergistic anti-tumor effect with PD-L1 blockade by inducing an immune response in ovarian cancer.
Journal of translational medicine
Meng J,Peng J,Feng J,Maurer J,Li X,Li Y,Yao S,Chu R,Pan X,Li J,Zhang T,Liu L,Zhang Q,Yuan Z,Bu H,Song K,Kong B
Published figure using IFN gamma monoclonal antibody (Product # 14-7319-81) in Flow Cytometry
Thu Oct 07 00:00:00 EDT 2021
Niraparib exhibits a synergistic anti-tumor effect with PD-L1 blockade by inducing an immune response in ovarian cancer.
Journal of translational medicine
Meng J,Peng J,Feng J,Maurer J,Li X,Li Y,Yao S,Chu R,Pan X,Li J,Zhang T,Liu L,Zhang Q,Yuan Z,Bu H,Song K,Kong B
Published figure using IFN gamma monoclonal antibody (Product # 14-7319-81) in Flow Cytometry
Thu Oct 07 00:00:00 EDT 2021
Lasting antibody and T cell responses to SARS-CoV-2 in COVID-19 patients three months after infection.
Nature communications
Jiang XL,Wang GL,Zhao XN,Yan FH,Yao L,Kou ZQ,Ji SX,Zhang XL,Li CB,Duan LJ,Li Y,Zhang YW,Duan Q,Wang TC,Li ET,Wei X,Wang QY,Wang XF,Sun WY,Gao YW,Kang DM,Zhang JY,Ma MJ
Published figure using IFN gamma monoclonal antibody (Product # 14-7319-81) in Flow Cytometry
Tue Feb 09 00:00:00 EST 2021
Identification of HLA-A2-Restricted Mutant Epitopes from Neoantigens of Esophageal Squamous Cell Carcinoma.
Vaccines
Wang Z,Ran L,Chen C,Shi R,Dong Y,Li Y,Zhou X,Qi Y,Zhu P,Gao Y,Wu Y
Published figure using IFN gamma monoclonal antibody (Product # 14-7319-81) in Flow Cytometry
Fri Oct 01 00:00:00 EDT 2021
NKD2 mediates stimulation-dependent ORAI1 trafficking to augment Ca2+ entry in T cells.
Cell reports
Wu B,Woo JS,Vila P,Jew M,Leung J,Sun Z,Srikanth S,Gwack Y
Published figure using IFN gamma monoclonal antibody (Product # 14-7319-81) in Flow Cytometry
Tue Aug 24 00:00:00 EDT 2021
Multipeptide stimulated PBMCs generate TEM/TCM for adoptive cell therapy in multiple myeloma.
Oncotarget
Vardam-Kaur T,Pathangey LB,McCormick DJ,Bergsagel PL,Cohen PA,Gendler SJ
Published figure using IFN gamma monoclonal antibody (Product # 14-7319-81) in Flow Cytometry
Tue Sep 28 00:00:00 EDT 2021
YY1 regulation by miR-124-3p promotes Th17 cell pathogenicity through interaction with T-bet in rheumatoid arthritis.
JCI insight
Lin J,Tang J,Lin J,He Y,Yu Z,Jiang R,Yang B,Ou Q
Published figure using IFN gamma monoclonal antibody (Product # 14-7319-81) in Flow Cytometry
Mon Nov 22 00:00:00 EST 2021
Rejuvenating Effector/Exhausted CAR T Cells to Stem Cell Memory-Like CAR T Cells By Resting Them in the Presence of CXCL12 and the NOTCH Ligand.
Cancer research communications
Ando M,Kondo T,Tomisato W,Ito M,Shichino S,Srirat T,Mise-Omata S,Nakagawara K,Yoshimura A
Published figure using IFN gamma monoclonal antibody (Product # 14-7319-81) in Flow Cytometry
Fri Oct 01 00:00:00 EDT 2021
Inhibition of Cholesterol Esterification Enzyme Enhances the Potency of Human Chimeric Antigen Receptor T Cells against Pancreatic Carcinoma.
Molecular therapy oncolytics
Zhao L,Liu Y,Zhao F,Jin Y,Feng J,Geng R,Sun J,Kang L,Yu L,Wei Y
Published figure using IFN gamma monoclonal antibody (Product # 14-7319-81) in Flow Cytometry
Fri Mar 27 00:00:00 EDT 2020
TIRC7 inhibits Th1 cells by upregulating the expression of CTLA‑4 and STAT3 in mice with acute graft‑versus‑host disease.
Oncology reports
Zhu F,Qiu T,Zhu S,Zhao K,Chen C,Qiao J,Pan B,Yan Z,Chen W,Liu Q,Wu Q,Cao J,Sang W,Zeng L,Sun H,Li Z,Xu K
Published figure using IFN gamma monoclonal antibody (Product # 14-7319-81) in Flow Cytometry
Wed Jul 01 00:00:00 EDT 2020
Differential Activation of Unconventional T Cells, Including iNKT Cells, in Alcohol-Related Liver Disease.
Alcoholism, clinical and experimental research
Marrero I,Maricic I,Morgan TR,Stolz AA,Schnabl B,Liu ZX,Tsukamoto H,Kumar V
14-7319-81 was used in Flow Cytometry to investigate the role of innate-like unconventional T cells in alcohol-related liver disease.
Fri May 01 00:00:00 EDT 2020
Immunoprophylactic and immunotherapeutic control of hormone receptor-positive breast cancer.
Nature communications
Buqué A,Bloy N,Perez-Lanzón M,Iribarren K,Humeau J,Pol JG,Levesque S,Mondragon L,Yamazaki T,Sato A,Aranda F,Durand S,Boissonnas A,Fucikova J,Senovilla L,Enot D,Hensler M,Kremer M,Stoll G,Hu Y,Massa C,Formenti SC,Seliger B,Elemento O,Spisek R,André F,Zitvogel L,Delaloge S,Kroemer G,Galluzzi L
Published figure using IFN gamma monoclonal antibody (Product # 14-7319-81) in Flow Cytometry
Thu Jul 30 00:00:00 EDT 2020
Single-cell transcriptomics identifies an effectorness gradient shaping the response of CD4+ T cells to cytokines.
Nature communications
Cano-Gamez E,Soskic B,Roumeliotis TI,So E,Smyth DJ,Baldrighi M,Willé D,Nakic N,Esparza-Gordillo J,Larminie CGC,Bronson PG,Tough DF,Rowan WC,Choudhary JS,Trynka G
Published figure using IFN gamma monoclonal antibody (Product # 14-7319-81) in Flow Cytometry
Tue Apr 14 00:00:00 EDT 2020
Ovarian granulosa cell tumor characterization identifies FOXL2 as an immunotherapeutic target.
JCI insight
Pierini S,Tanyi JL,Simpkins F,George E,Uribe-Herranz M,Drapkin R,Burger R,Morgan MA,Facciabene A
Published figure using IFN gamma monoclonal antibody (Product # 14-7319-81) in Flow Cytometry
Thu Aug 20 00:00:00 EDT 2020
A Method of Assessment of Human Natural Killer Cell Phenotype and Function in Whole Blood.
Frontiers in immunology
Market M,Tennakoon G,Ng J,Scaffidi M,de Souza CT,Kennedy MA,Auer RC
Published figure using IFN gamma monoclonal antibody (Product # 14-7319-81) in ELISA
Mon Mar 29 00:00:00 EDT 2021
A Method of Assessment of Human Natural Killer Cell Phenotype and Function in Whole Blood.
Frontiers in immunology
Market M,Tennakoon G,Ng J,Scaffidi M,de Souza CT,Kennedy MA,Auer RC
Published figure using IFN gamma monoclonal antibody (Product # 14-7319-81) in ELISA
Mon Mar 29 00:00:00 EDT 2021
Human Tumor-Infiltrating MAIT Cells Display Hallmarks of Bacterial Antigen Recognition in Colorectal Cancer.
Cell reports. Medicine
Li S,Simoni Y,Becht E,Loh CY,Li N,Lachance D,Koo SL,Lim TP,Tan EKW,Mathew R,Nguyen A,Golovato J,Berkson JD,Prlic M,Lee B,Minot SS,Nagarajan N,Dey N,Tan DSW,Tan IB,Newell EW
Published figure using IFN gamma monoclonal antibody (Product # 14-7319-81) in Flow Cytometry
Tue Jun 23 00:00:00 EDT 2020
NPM1 upregulates the transcription of PD-L1 and suppresses T cell activity in triple-negative breast cancer.
Nature communications
Qin G,Wang X,Ye S,Li Y,Chen M,Wang S,Qin T,Zhang C,Li Y,Long Q,Hu H,Shi D,Li J,Zhang K,Zhai Q,Tang Y,Kang T,Lan P,Xie F,Lu J,Deng W
Published figure using IFN gamma monoclonal antibody (Product # 14-7319-81) in Flow Cytometry
Fri Apr 03 00:00:00 EDT 2020
CAIX Regulates GBM Motility and TAM Adhesion and Polarization through EGFR/STAT3 under Hypoxic Conditions.
International journal of molecular sciences
Huang BR,Liu YS,Lai SW,Lin HJ,Shen CK,Yang LY,Lu DY
Published figure using IFN gamma monoclonal antibody (Product # 14-7319-81) in Flow Cytometry
Fri Aug 14 00:00:00 EDT 2020
Duration of Cellular and Humoral Responses after Quadrivalent Human Papillomavirus Vaccination in Healthy Female Adults with or without Prior Type 16 and/or 18 Exposure.
Vaccines
Lai L,Ault K,Rouphael N,Beck A,Domjahn B,Xu Y,Anderson EJ,Cheng A,Nakamura A,Hoagland RJ,Kelley C,Edupuganti S,Mask K,Nesin M,Unger ER,Panicker G,David H,Mulligan MJ
Published figure using IFN gamma monoclonal antibody (Product # 14-7319-81) in Flow Cytometry
Tue Jun 30 00:00:00 EDT 2020
Th2 Biased Immunity With Altered B Cell Profiles in Circulation of Patients With Sporotrichosis Caused by Sporothrix globosa.
Frontiers in immunology
Zu J,Yao L,Song Y,Cui Y,Guan M,Chen R,Zhen Y,Li S
Published figure using IFN gamma monoclonal antibody (Product # 14-7319-81) in Flow Cytometry
Mon May 31 00:00:00 EDT 2021
Engineered triple inhibitory receptor resistance improves anti-tumor CAR-T cell performance via CD56.
Nature communications
Zou F,Lu L,Liu J,Xia B,Zhang W,Hu Q,Liu W,Zhang Y,Lin Y,Jing S,Huang M,Huang B,Liu B,Zhang H
Published figure using IFN gamma monoclonal antibody (Product # 14-7319-81) in Flow Cytometry
Wed Sep 11 00:00:00 EDT 2019
TIGIT signaling restores suppressor function of Th1 Tregs.
JCI insight
Lucca LE,Axisa PP,Singer ER,Nolan NM,Dominguez-Villar M,Hafler DA
14-7319 was used in Flow cytometry/Cell sorting to demonstrate an important role for TIGIT in controlling the functional stability of Tregs through repression of Akt, suggesting that the TIGIT pathway could be targeted for immunomodulatory therapies in human autoimmune disorders.
Thu Feb 07 00:00:00 EST 2019
Cytomegalovirus-Driven Adaption of Natural Killer Cells in NKG2Cnull Human Immunodeficiency Virus-Infected Individuals.
Viruses
Comeau EM,Holder KA,Fudge NJ,Grant MD
14-7319-81 was used in Flow Cytometry to investigate whether the loss of NKG2C expression affects natural killer cell acquisition of superior antibody-dependent function in the diminished resistance to HIV infection seen with this loss.
Sat Mar 09 00:00:00 EST 2019
Exposure to wild-type AAV drives distinct capsid immunity profiles in humans.
The Journal of clinical investigation
Kuranda K,Jean-Alphonse P,Leborgne C,Hardet R,Collaud F,Marmier S,Costa Verdera H,Ronzitti G,Veron P,Mingozzi F
Published figure using IFN gamma monoclonal antibody (Product # 14-7319-81) in Flow Cytometry
Mon Dec 03 00:00:00 EST 2018
Optimized RNP transfection for highly efficient CRISPR/Cas9-mediated gene knockout in primary T cells.
The Journal of experimental medicine
Seki A,Rutz S
14-7319 was used in Flow cytometry/Cell sorting to develop an optimised approach for Cas9/RNP transfection of primary mouse and human T cells without TCR stimulation, that results in near complete loss of target gene expression at the population level.
Mon Mar 05 00:00:00 EST 2018
CD4 T cell loss and Th2 and Th17 bias are associated with the severity of severe fever with thrombocytopenia syndrome (SFTS).
Clinical immunology (Orlando, Fla.)
Li MM,Zhang WJ,Weng XF,Li MY,Liu J,Xiong Y,Xiong SE,Zou CC,Wang H,Lu MJ,Yang DL,Peng C,Zheng X
Published figure using IFN gamma monoclonal antibody (Product # 14-7319-81) in Flow Cytometry
Mon Oct 01 00:00:00 EDT 2018
RKIP mediates autoimmune inflammation by positively regulating IL-17R signaling.
EMBO reports
Lin W,Wang N,Zhou K,Su F,Jiang Y,Shou J,Liu H,Ma C,Qian Y,Wang K,Wang X
Published figure using IFN gamma monoclonal antibody (Product # 14-7319-81) in Flow Cytometry
Fri Jun 01 00:00:00 EDT 2018
TCF1 expression marks self-renewing human CD8+ T cells.
Blood advances
Kratchmarov R,Magun AM,Reiner SL
14-7319 was used in Flow cytometry/Cell sorting to show analyses of TCF1 expression from patient specimens may become a useful biomarker for adaptive immune function in response to vaccination, infection, autoimmunity, and cancer.
Tue Jul 24 00:00:00 EDT 2018
CXCL4 is a novel inducer of human Th17 cells and correlates with IL-17 and IL-22 in psoriatic arthritis.
European journal of immunology
Affandi AJ,Silva-Cardoso SC,Garcia S,Leijten EFA,van Kempen TS,Marut W,van Roon JAG,Radstake TRDJ
Published figure using IFN gamma monoclonal antibody (Product # 14-7319-81) in Flow Cytometry
Thu Mar 01 00:00:00 EST 2018
Accumulation of T-helper 22 cells, interleukin-22 and myeloid-derived suppressor cells promotes gastric cancer progression in elderly patients.
Oncology letters
Chen X,Wang Y,Wang J,Wen J,Jia X,Wang X,Zhang H
Published figure using IFN gamma monoclonal antibody (Product # 14-7319-81) in Flow Cytometry
Sun Jul 01 00:00:00 EDT 2018
Chloroquine modulates antitumor immune response by resetting tumor-associated macrophages toward M1 phenotype.
Nature communications
Chen D,Xie J,Fiskesund R,Dong W,Liang X,Lv J,Jin X,Liu J,Mo S,Zhang T,Cheng F,Zhou Y,Zhang H,Tang K,Ma J,Liu Y,Huang B
Published figure using IFN gamma monoclonal antibody (Product # 14-7319-81) in Flow Cytometry
Wed Feb 28 00:00:00 EST 2018
Human in vivo-generated monocyte-derived dendritic cells and macrophages cross-present antigens through a vacuolar pathway.
Nature communications
Tang-Huau TL,Gueguen P,Goudot C,Durand M,Bohec M,Baulande S,Pasquier B,Amigorena S,Segura E
Published figure using IFN gamma monoclonal antibody (Product # 14-7319-81) in Flow Cytometry
Mon Jul 02 00:00:00 EDT 2018
The Phenotype of Monocytes in Anterior Uveitis Depends on the HLA-B27 Status.
Frontiers in immunology
Kasper M,Walscheid K,Laffer B,Bauer D,Busch M,Wildschütz L,Wang B,Loser K,Vogl T,Grajewski RS,Langmann T,Heiligenhaus A
14-7319 was used in Flow cytometry/Cell sorting to show the differentially regulated response of both innate and adaptive immune cells in the blood may be related to the clinically distinct characteristics of the two different uveitis entities.
Mon Sep 16 00:00:00 EDT 2019
Natural Killer Cells Adapt to Cytomegalovirus Along a Functionally Static Phenotypic Spectrum in Human Immunodeficiency Virus Infection.
Frontiers in immunology
Holder KA,Lajoie J,Grant MD
14-7319 was used in Flow cytometry/Cell sorting to investigate natural killer cell adaptation to human cytomegalovirus along a functionally static phenotypic spectrum in Human Immunodeficiency Virus infection.
Mon Sep 30 00:00:00 EDT 2019
Fibroblast-adapted human CMV vaccines elicit predominantly conventional CD8 T cell responses in humans.
The Journal of experimental medicine
Murray SE,Nesterenko PA,Vanarsdall AL,Munks MW,Smart SM,Veziroglu EM,Sagario LC,Lee R,Claas FHJ,Doxiadis IIN,McVoy MA,Adler SP,Hill AB
14-7319 was used in Flow cytometry/Cell sorting to describe the CD8 T cell responses of human subjects vaccinated with two fibroblast-adapted HCMV vaccines.
Mon Jul 03 00:00:00 EDT 2017
Impact of Enhancer of Zeste Homolog 2 on T Helper Cell-Mediated Allergic Rhinitis.
Frontiers in immunology
Hou TY,Chen MR,Chou YC,Kan PC,Tsai YT,Cha TL
Published figure using IFN gamma monoclonal antibody (Product # 14-7319-81) in Flow Cytometry
Wed Nov 20 00:00:00 EST 2019
Ebola virus glycoprotein directly triggers T lymphocyte death despite of the lack of infection.
PLoS pathogens
Iampietro M,Younan P,Nishida A,Dutta M,Lubaki NM,Santos RI,Koup RA,Katze MG,Bukreyev A
Published figure using IFN gamma monoclonal antibody (Product # 14-7319-81) in Flow Cytometry
Mon May 01 00:00:00 EDT 2017
Dominant immunosuppression of dendritic cell function by prostate-cancer-derived exosomes.
Journal of extracellular vesicles
Salimu J,Webber J,Gurney M,Al-Taei S,Clayton A,Tabi Z
Published figure using IFN gamma monoclonal antibody (Product # 14-7319-81) in Flow Cytometry
Wed Sep 30 00:00:00 EDT 2020
Single-cell profiling reveals GPCR heterogeneity and functional patterning during neuroinflammation.
JCI insight
Tischner D,Grimm M,Kaur H,Staudenraus D,Carvalho J,Looso M,Günther S,Wanke F,Moos S,Siller N,Breuer J,Schwab N,Zipp F,Waisman A,Kurschus FC,Offermanns S,Wettschureck N
Published figure using IFN gamma monoclonal antibody (Product # 14-7319-81) in Flow Cytometry
Thu Aug 03 00:00:00 EDT 2017
Surrogate in vitro activation of innate immunity synergizes with interleukin-7 to unleash rapid antigen-driven outgrowth of CD4+ and CD8+ human peripheral blood T-cells naturally recognizing MUC1, HER2/neu and other tumor-associated antigens.
Oncotarget
Pathangey LB,McCurry DB,Gendler SJ,Dominguez AL,Gorman JE,Pathangey G,Mihalik LA,Dang Y,Disis ML,Cohen PA
Published figure using IFN gamma monoclonal antibody (Product # 14-7319-81) in Flow Cytometry
Tue Feb 14 00:00:00 EST 2017
Commensal-Specific CD4(+) Cells From Patients With Crohn's Disease Have a T-Helper 17 Inflammatory Profile.
Gastroenterology
Calderón-Gómez E,Bassolas-Molina H,Mora-Buch R,Dotti I,Planell N,Esteller M,Gallego M,Martí M,Garcia-Martín C,Martínez-Torró C,Ordás I,Singh S,Panés J,Benítez-Ribas D,Salas A
14-7319 was used in Flow cytometry/Cell sorting to investigate the antigen-specificity of T cells isolated from patients with Crohn's disease.
Thu Sep 01 00:00:00 EDT 2016
T regulatory cell chemokine production mediates pathogenic T cell attraction and suppression.
The Journal of clinical investigation
Patterson SJ,Pesenacker AM,Wang AY,Gillies J,Mojibian M,Morishita K,Tan R,Kieffer TJ,Verchere CB,Panagiotopoulos C,Levings MK
14-7319 was used in Flow cytometry/Cell sorting to investigate how Treg-to-target proximity is controlled.
Tue Mar 01 00:00:00 EST 2016
CD28 family of receptors on T cells in chronic HBV infection: Expression characteristics, clinical significance and correlations with PD-1 blockade.
Molecular medicine reports
Tang ZS,Hao YH,Zhang EJ,Xu CL,Zhou Y,Zheng X,Yang DL
Published figure using IFN gamma monoclonal antibody (Product # 14-7319-81) in Flow Cytometry
Mon Aug 01 00:00:00 EDT 2016
Heterologous Immunity between Adenoviruses and Hepatitis C Virus: A New Paradigm in HCV Immunity and Vaccines.
PloS one
Singh S,Vedi S,Samrat SK,Li W,Kumar R,Agrawal B
Published figure using IFN gamma monoclonal antibody (Product # 14-7319-81) in Flow Cytometry
Tue Jul 19 00:00:00 EDT 2016
Peritoneal carcinomatosis of colorectal cancer is characterized by structural and functional reorganization of the tumor microenvironment inducing senescence and proliferation arrest in cancer cells.
Oncoimmunology
Seebauer CT,Brunner S,Glockzin G,Piso P,Ruemmele P,Schlitt HJ,Geissler EK,Fichtner-Feigl S,Kesselring R
Published figure using IFN gamma monoclonal antibody (Product # 14-7319-81) in Flow Cytometry
Tue Jan 12 00:00:00 EST 2021
The RNA-Binding Protein, Polypyrimidine Tract-Binding Protein 1 (PTBP1) Is a Key Regulator of CD4 T Cell Activation.
PloS one
La Porta J,Matus-Nicodemos R,Valentín-Acevedo A,Covey LR
14731981 was used in flow cytometry to investigate the role of polypyrimidine tract-binding protein 1 in CD4 T cell activation
Thu Jul 27 00:00:00 EDT 2017
Functional dichotomy of Vδ2 γδ T cells in chronic hepatitis C virus infections: role in cytotoxicity but not for IFN-γ production.
Scientific reports
Yin W,Tong S,Zhang Q,Shao J,Liu Q,Peng H,Hu H,Peng M,Hu P,Ren H,Tian Z,Zhang D
14-7319 was used in Flow cytometry/Cell sorting to analyse the phenotype and function of Vδ2 T cells in HCV-infected patients compared to healthy controls.
Thu May 19 00:00:00 EDT 2016
COPA mutations impair ER-Golgi transport and cause hereditary autoimmune-mediated lung disease and arthritis.
Nature genetics
Watkin LB,Jessen B,Wiszniewski W,Vece TJ,Jan M,Sha Y,Thamsen M,Santos-Cortez RL,Lee K,Gambin T,Forbes LR,Law CS,Stray-Pedersen A,Cheng MH,Mace EM,Anderson MS,Liu D,Tang LF,Nicholas SK,Nahmod K,Makedonas G,Canter DL,Kwok PY,Hicks J,Jones KD,Penney S,Jhangiani SN,Rosenblum MD,Dell SD,Waterfield MR,Papa FR,Muzny DM,Zaitlen N,Leal SM,Gonzaga-Jauregui C,Boerwinkle E,Eissa NT,Gibbs RA,Lupski JR,Orange JS,Shum AK
14-7319 was used in Flow cytometry/Cell sorting to investigate how a vesicular transport protein and a syndrome of autoimmunity manifested by lung and joint disease have an unexpected molecular link.
Mon Jun 01 00:00:00 EDT 2015
A SnoRNA-derived piRNA interacts with human interleukin-4 pre-mRNA and induces its decay in nuclear exosomes.
Nucleic acids research
Zhong F,Zhou N,Wu K,Guo Y,Tan W,Zhang H,Zhang X,Geng G,Pan T,Luo H,Zhang Y,Xu Z,Liu J,Liu B,Gao W,Liu C,Ren L,Li J,Zhou J,Zhang H
Published figure using IFN gamma monoclonal antibody (Product # 14-7319-81) in Flow Cytometry
Wed Dec 02 00:00:00 EST 2015
Brief Report: IFIH1 Mutation Causes Systemic Lupus Erythematosus With Selective IgA Deficiency.
Arthritis & rheumatology (Hoboken, N.J.)
Van Eyck L,De Somer L,Pombal D,Bornschein S,Frans G,Humblet-Baron S,Moens L,de Zegher F,Bossuyt X,Wouters C,Liston A
14-7319 was used in Flow cytometry/Cell sorting to determine the underlying genetic defect responsible for severe early onset and refractory systemic lupus erythematosus.
Mon Jun 01 00:00:00 EDT 2015
The Immunology of a Healing Response in Cutaneous Leishmaniasis Treated with Localized Heat or Systemic Antimonial Therapy.
PLoS neglected tropical diseases
Lakhal-Naouar I,Slike BM,Aronson NE,Marovich MA
14-7319 was used in Flow cytometry/Cell sorting to discover that healing from cutaneous Leishmaniasis is dynamic, with differing circulating lymphocyte populations and subsets of T, NK and NKT-like cells.
Mon Mar 28 00:00:00 EDT 2016
Safety and immunogenicity of a candidate tuberculosis vaccine MVA85A delivered by aerosol in BCG-vaccinated healthy adults: a phase 1, double-blind, randomised controlled trial.
The Lancet. Infectious diseases
Satti I,Meyer J,Harris SA,Manjaly Thomas ZR,Griffiths K,Antrobus RD,Rowland R,Ramon RL,Smith M,Sheehan S,Bettinson H,McShane H
14-7319 was used in Flow cytometry/Cell sorting to compare the safety and immunogenicity of aerosol-administered and intradermally administered MVA85A, a candidate vaccine against tuberculosis.
Wed Oct 01 00:00:00 EDT 2014
Signaling lymphocytic activation molecule (SLAM)/SLAM-associated protein pathway regulates human B-cell tolerance.
The Journal of allergy and clinical immunology
Menard L,Cantaert T,Chamberlain N,Tangye SG,Riminton S,Church JA,Klion A,Cunningham-Rundles C,Nichols KE,Meffre E
14-7319 was used in Flow cytometry/Cell sorting to show SLAM-associated protein expression is necessary for the counterselection of developing autoreactive B cells and prevents their T cell-dependent accumulation in the periphery.
Tue Apr 01 00:00:00 EDT 2014
Multifactorial T-cell hypofunction that is reversible can limit the efficacy of chimeric antigen receptor-transduced human T cells in solid tumors.
Clinical cancer research : an official journal of the American Association for Cancer Research
Moon EK,Wang LC,Dolfi DV,Wilson CB,Ranganathan R,Sun J,Kapoor V,Scholler J,Puré E,Milone MC,June CH,Riley JL,Wherry EJ,Albelda SM
Published figure using IFN gamma monoclonal antibody (Product # 14-7319-81) in Flow Cytometry
Fri Aug 15 00:00:00 EDT 2014
T cell inactivation by poxviral B22 family proteins increases viral virulence.
PLoS pathogens
Alzhanova D,Hammarlund E,Reed J,Meermeier E,Rawlings S,Ray CA,Edwards DM,Bimber B,Legasse A,Planer S,Sprague J,Axthelm MK,Pickup DJ,Lewinsohn DM,Gold MC,Wong SW,Sacha JB,Slifka MK,Früh K
14-7319 was used in Flow cytometry/Cell sorting to conclude that B22 family proteins cause viral virulence by suppressing T cell control of viral dissemination.
Thu May 01 00:00:00 EDT 2014
T cell inactivation by poxviral B22 family proteins increases viral virulence.
PLoS pathogens
Alzhanova D,Hammarlund E,Reed J,Meermeier E,Rawlings S,Ray CA,Edwards DM,Bimber B,Legasse A,Planer S,Sprague J,Axthelm MK,Pickup DJ,Lewinsohn DM,Gold MC,Wong SW,Sacha JB,Slifka MK,Früh K
14-7319 was used in Flow cytometry/Cell sorting to conclude that B22 family proteins cause viral virulence by suppressing T cell control of viral dissemination.
Thu May 01 00:00:00 EDT 2014
Failure to upregulate cell surface PD-1 is associated with dysregulated stimulation of T cells by TGN1412-like CD28 superagonist.
mAbs
Thaventhiran T,Alhumeed N,Yeang HX,Sethu S,Downey JS,Alghanem AF,Olayanju A,Smith EL,Cross MJ,Webb SD,Williams DP,Bristow A,Ball C,Stebbings R,Sathish JG
Published figure using IFN gamma monoclonal antibody (Product # 14-7319-81) in Flow Cytometry
Mon Aug 24 00:00:00 EDT 2015
The impact of IFN-γ receptor on SLPI expression in active tuberculosis: association with disease severity.
The American journal of pathology
Tateosian NL,Pasquinelli V,Hernández Del Pino RE,Ambrosi N,Guerrieri D,Pedraza-Sánchez S,Santucci N,D'Attilio L,Pellegrini J,Araujo-Solis MA,Musella RM,Palmero DJ,Hernandez-Pando R,Garcia VE,Chuluyan HE
14-7319 was used in an ELISA assay to show that Mycobacterium tuberculosis-induced IFN-γ down-modulated SLPI levels by signalling through IFN-γR in healthy donors.
Thu May 01 00:00:00 EDT 2014
Hemin controls T cell polarization in sickle cell alloimmunization.
Journal of immunology (Baltimore, Md. : 1950)
Zhong H,Bao W,Friedman D,Yazdanbakhsh K
14-7319 was used in Flow cytometry/Cell sorting to investigate whether monocyte control of T cell polarisation differs between alloimmunised and non-alloimmunised sickle cell disease patients.
Tue Jul 01 00:00:00 EDT 2014
Microbe-specific unconventional T cells induce human neutrophil differentiation into antigen cross-presenting cells.
Journal of immunology (Baltimore, Md. : 1950)
Davey MS,Morgan MP,Liuzzi AR,Tyler CJ,Khan MWA,Szakmany T,Hall JE,Moser B,Eberl M
14-7319 was used in Immunofluorescence to investigate how unconventional T cells represent key controllers of neutrophil-driven innate and adaptive responses to a broad range of pathogens.
Wed Oct 01 00:00:00 EDT 2014
Akt-dependent enhanced migratory capacity of Th17 cells from children with lupus nephritis.
Journal of immunology (Baltimore, Md. : 1950)
Kshirsagar S,Riedl M,Billing H,Tönshoff B,Thangavadivel S,Steuber C,Staude H,Wechselberger G,Edelbauer M
14-7319 was used in Flow cytometry/Cell sorting to determine that therapeutic modulation of Akt activity could suppress chronic inflammatory processes in children with lupus.
Sat Nov 15 00:00:00 EST 2014
Tumor-derived exosomes promote tumor progression and T-cell dysfunction through the regulation of enriched exosomal microRNAs in human nasopharyngeal carcinoma.
Oncotarget
Ye SB,Li ZL,Luo DH,Huang BJ,Chen YS,Zhang XS,Cui J,Zeng YX,Li J
Published figure using IFN gamma monoclonal antibody (Product # 14-7319-81) in Flow Cytometry
Wed Jul 30 00:00:00 EDT 2014
CD40L expression permits CD8+ T cells to execute immunologic helper functions.
Blood
Frentsch M,Stark R,Matzmohr N,Meier S,Durlanik S,Schulz AR,Stervbo U,Jürchott K,Gebhardt F,Heine G,Reuter MA,Betts MR,Busch D,Thiel A
14-7319 was used in Flow cytometry/Cell sorting to investigate the role of CD40L in CD8+ T cells, showing that expression permits CD8+ T cells to execute immunologic helper functions.
Thu Jul 18 00:00:00 EDT 2013
Peptide vaccination in Montanide adjuvant induces and GM-CSF increases CXCR3 and cutaneous lymphocyte antigen expression by tumor antigen-specific CD8 T cells.
Cancer immunology research
Clancy-Thompson E,King LK,Nunnley LD,Mullins IM,Slingluff CL,Mullins DW
14-7319 was used in Flow cytometry/Cell sorting to assess the melanoma-infiltrating capability of CD8 T cells in the context of a clinical trial of a melanoma-specific peptide vaccine.
Fri Nov 01 00:00:00 EDT 2013
Somatostatin expression in human hair follicles and its potential role in immune privilege.
The Journal of investigative dermatology
Breitkopf T,Lo BK,Leung G,Wang E,Yu M,Carr N,Zloty D,Cowan B,Shapiro J,McElwee KJ
14-7319 was used in Flow cytometry/Cell sorting to demonstrate and quantify the functional immune privilege properties apparent in human hair follicles.
Mon Jul 01 00:00:00 EDT 2013
Phase I trial of a multi-epitope-pulsed dendritic cell vaccine for patients with newly diagnosed glioblastoma.
Cancer immunology, immunotherapy : CII
Phuphanich S,Wheeler CJ,Rudnick JD,Mazer M,Wang H,Nuño MA,Richardson JE,Fan X,Ji J,Chu RM,Bender JG,Hawkins ES,Patil CG,Black KL,Yu JS
14-7319 was used in Flow cytometry/Cell sorting to study how expression of four ICT-107 targeted antigens in the pre-vaccine tumors correlated with prolonged overall survival and PFS in ND-GBM patients.
Tue Jan 01 00:00:00 EST 2013
Elevated frequencies of circulating Th22 cell in addition to Th17 cell and Th17/Th1 cell in patients with acute coronary syndrome.
PloS one
Zhang L,Wang T,Wang XQ,Du RZ,Zhang KN,Liu XG,Ma DX,Yu S,Su GH,Li ZH,Guan YQ,Du NL
Published figure using IFN gamma monoclonal antibody (Product # 14-7319-81) in Flow Cytometry
Mon Aug 18 00:00:00 EDT 2014
CD137L-stimulated dendritic cells are more potent than conventional dendritic cells at eliciting cytotoxic T-cell responses.
Oncoimmunology
Harfuddin Z,Kwajah S,Chong Nyi Sim A,Macary PA,Schwarz H
14-7319 was used in an ELISA assay to develop improved dendritic cell-based immunotherapy using CD137 signalling.
Fri Nov 01 00:00:00 EDT 2013
Increased frequencies of Th22 cells as well as Th17 cells in the peripheral blood of patients with ankylosing spondylitis and rheumatoid arthritis.
PloS one
Zhang L,Li YG,Li YH,Qi L,Liu XG,Yuan CZ,Hu NW,Ma DX,Li ZF,Yang Q,Li W,Li JM
Published figure using IFN gamma monoclonal antibody (Product # 14-7319-81) in Flow Cytometry
Fri Jul 27 00:00:00 EDT 2012
Characterization of interleukin-17-producing regulatory T cells in inflamed intestinal mucosa from patients with inflammatory bowel diseases.
Gastroenterology
Hovhannisyan Z,Treatman J,Littman DR,Mayer L
14-7319 was used in Flow cytometry/Cell sorting to investigate how Th17 and Tregs interact under chronic inflammatory conditions.
Tue Mar 01 00:00:00 EST 2011
CD137 ligand signaling induces human monocyte to dendritic cell differentiation.
European journal of immunology
Kwajah M M S,Schwarz H
14-7319 was used in Flow cytometry to deduce that peripheral monocytes can be differentiated into mature inflammatory dendritic cells by the single factor CD137.
Thu Jul 01 00:00:00 EDT 2010
Cowpox virus inhibits the transporter associated with antigen processing to evade T cell recognition.
Cell host & microbe
Alzhanova D,Edwards DM,Hammarlund E,Scholz IG,Horst D,Wagner MJ,Upton C,Wiertz EJ,Slifka MK,Früh K
14-7319 was used in Flow cytometry/Cell sorting to identify cowpox virus proteins that mediate T cell evasion in multiple hosts.
Thu Nov 19 00:00:00 EST 2009
A role for human skin-resident T cells in wound healing.
The Journal of experimental medicine
Toulon A,Breton L,Taylor KR,Tenenhaus M,Bhavsar D,Lanigan C,Rudolph R,Jameson J,Havran WL
14-7319 was used in Flow cytometry/Cell sorting to demonstrate that human epidermal T cells are able to produce IGF-1 upon activation to promote wound healing.
Mon Apr 13 00:00:00 EDT 2009
Selective reduction of graft-versus-host disease-mediating human T cells by ex vivo treatment with soluble Fas ligand.
Journal of immunology (Baltimore, Md. : 1950)
Bohana-Kashtan O,Morisot S,Hildreth R,Brayton C,Levitsky HI,Civin CI
14-7319 was used in Flow cytometry/Cell sorting to investigate T cell ex vivo treatment with soluble Fas ligand as a mechanism to selectively reduce graft-versus-host-disease mediating cells.
Wed Jul 01 00:00:00 EDT 2009
Transfer of tolerance to collagen type V suppresses T-helper-cell-17 lymphocyte-mediated acute lung transplant rejection.
Transplantation
Braun RK,Molitor-Dart M,Wigfield C,Xiang Z,Fain SB,Jankowska-Gan E,Seroogy CM,Burlingham WJ,Wilkes DS,Brand DD,Torrealba J,Love RB
14-7319 was used in Neutralization experiments to test whether Tregs from tolerant rats could suppress Th17-mediated rejection in the syngeneic model of lung transplantation.
Sun Dec 27 00:00:00 EST 2009