Product References
Co-expression of IL-21-Enhanced NKG2D CAR-NK cell therapy for lung cancer.
BMC cancer
Zhang Y,Zhang C,He M,Xing W,Hou R,Zhang H
Published figure using IFN gamma monoclonal antibody (Product # 17-7319-41) in Flow Cytometry
Tue Jan 23 00:00:00 UTC 2024
Low level of ARID1A contributes to adaptive immune resistance and sensitizes triple-negative breast cancer to immune checkpoint inhibitors.
Cancer communications (London, England)
Chen XY,Li B,Wang Y,Jin J,Yang Y,Huang LH,Yang MD,Zhang J,Wang BY,Shao ZM,Ni T,Huang SL,Hu XC,Tao ZH
Published figure using IFN gamma monoclonal antibody (Product # 17-7319-41) in Flow Cytometry
Fri Sep 01 00:00:00 UTC 2023
Nanobody-derived bispecific CAR-T cell therapy enhances the anti-tumor efficacy of T cell lymphoma treatment.
Molecular therapy oncolytics
Xia B,Lin K,Wang X,Chen F,Zhou M,Li Y,Lin Y,Qiao Y,Li R,Zhang W,He X,Zou F,Li L,Lu L,Chen C,Li W,Zhang H,Liu B
Published figure using IFN gamma monoclonal antibody (Product # 17-7319-41) in Flow Cytometry
Thu Sep 21 00:00:00 UTC 2023
A Ferritin Nanoparticle-Based Zika Virus Vaccine Candidate Induces Robust Humoral and Cellular Immune Responses and Protects Mice from Lethal Virus Challenge.
Vaccines
Pattnaik A,Sahoo BR,Struble LR,Borgstahl GEO,Zhou Y,Franco R,Barletta RG,Osorio FA,Petro TM,Pattnaik AK
Published figure using IFN gamma monoclonal antibody (Product # 17-7319-41) in Flow Cytometry
Mon Apr 10 00:00:00 UTC 2023
Dynamics of humoral and cellular response to three doses of anti-SARS-CoV-2 BNT162b2 vaccine in patients with hematological malignancies and older subjects.
Frontiers in immunology
Laquintana V,Mottini C,Marchesi F,Marcozzi B,Terrenato I,Sperandio E,de Latouliere L,Carrieri F,Pimpinelli F,Pontone M,Pellini R,Campo F,Conti L,Accetta C,Mandoj C,Petrone F,Di Bella O,Vujovic B,Morrone A,Compagnone M,Principato E,Pinto E,Papa E,Falcucci P,La Malfa A,Pallocca M,De Marco F,Piaggio G,Ciliberto G,Mengarelli A,di Martino S
Published figure using IFN gamma monoclonal antibody (Product # 17-7319-41) in Flow Cytometry
Wed Feb 14 00:00:00 UTC 2024
Soluble NKG2D ligands impair CD8+ T cell antitumor function dependent of NKG2D downregulation in neuroblastoma.
Oncology letters
Zhang Y,Luo F,Dong K
Published figure using IFN gamma monoclonal antibody (Product # 17-7319-41) in Flow Cytometry
Sat Jul 01 00:00:00 UTC 2023
R274X-mutated Phf6 increased the self-renewal and skewed T cell differentiation of hematopoietic stem cells.
iScience
Lan Y,Yuan S,Guo T,Hou S,Zhao F,Yang W,Cao Y,Chu Y,Jiang E,Yuan W,Wang X
Published figure using IFN gamma monoclonal antibody (Product # 17-7319-41) in Flow Cytometry
Fri Jun 16 00:00:00 UTC 2023
Distinct SARS-CoV-2 specific NLRP3 and IL-1β responses in T cells of aging patients during acute COVID-19 infection.
Frontiers in immunology
Mahalingam SS,Jayaraman S,Arunkumar A,Dudley HM,Anthony DD,Shive CL,Jacobson JM,Pandiyan P
Published figure using IFN gamma monoclonal antibody (Product # 17-7319-41) in Flow Cytometry
Thu Oct 12 00:00:00 UTC 2023
Purinergic receptor P2Y12 boosts autoimmune hepatitis through hexokinase 2-dependent glycolysis in T cells.
International journal of biological sciences
Zhuang W,Liu X,Liu G,Lv J,Qin H,Wang C,Xie L,Saimaier K,Han S,Shi C,Hua Q,Zhang R,Du C
Published figure using IFN gamma monoclonal antibody (Product # 17-7319-41) in Flow Cytometry
Thu Aug 03 00:00:00 UTC 2023
CD8+ T cell-Dependent Remodeling of the Tumor Microenvironment Overcomes Chemoresistance.
Cancer immunology research
Lao L,Zeng W,Huang P,Chen H,Jia Z,Wang P,Huang D,Chen J,Nie Y,Yang L,Wu W,Liu J
Published figure using IFN gamma monoclonal antibody (Product # 17-7319-41) in Flow Cytometry
Wed Mar 01 00:00:00 UTC 2023
Rapamycin improves Graves' orbitopathy by suppressing CD4+ cytotoxic T lymphocytes.
JCI insight
Zhang M,Chong KK,Chen ZY,Guo H,Liu YF,Kang YY,Li YJ,Shi TT,Lai KK,He MQ,Ye K,Kahaly GJ,Shi BY,Wang Y
Published figure using IFN gamma monoclonal antibody (Product # 17-7319-41) in Flow Cytometry
Wed Feb 08 00:00:00 UTC 2023
HERC2 promotes inflammation-driven cancer stemness and immune evasion in hepatocellular carcinoma by activating STAT3 pathway.
Journal of experimental & clinical cancer research : CR
Liu Y,Xu Q,Deng F,Zheng Z,Luo J,Wang P,Zhou J,Lu X,Zhang L,Chen Z,Zhang Q,Chen Q,Zuo D
Published figure using IFN gamma monoclonal antibody (Product # 17-7319-41) in Flow Cytometry
Wed Feb 01 00:00:00 UTC 2023
GPR116 receptor regulates the antitumor function of NK cells via Gαq/HIF1α/NF-κB signaling pathway as a potential immune checkpoint.
Cell & bioscience
Guo D,Jin C,Gao Y,Lin H,Zhang L,Zhou Y,Yao J,Duan Y,Ren Y,Hui X,Ge Y,Yang R,Jiang W
Published figure using IFN gamma monoclonal antibody (Product # 17-7319-41) in Flow Cytometry
Thu Mar 09 00:00:00 UTC 2023
A Novel Homozygous Stop Mutation in IL23R Causes Mendelian Susceptibility to Mycobacterial Disease.
Journal of clinical immunology
Staels F,Lorenzetti F,De Keukeleere K,Willemsen M,Gerbaux M,Neumann J,Tousseyn T,Pasciuto E,De Munter P,Bossuyt X,Gijsbers R,Liston A,Humblet-Baron S,Schrijvers R
Published figure using IFN gamma monoclonal antibody (Product # 17-7319-41) in Flow Cytometry
Tue Nov 01 00:00:00 UTC 2022
GM-CSF-activated human dendritic cells promote type 1 T follicular helper cell polarization in a CD40-dependent manner.
Journal of cell science
Korniotis S,Saichi M,Trichot C,Hoffmann C,Amblard E,Viguier A,Grondin S,Noel F,Mattoo H,Soumelis V
Published figure using IFN gamma monoclonal antibody (Product # 17-7319-41) in Flow Cytometry
Tue Nov 01 00:00:00 UTC 2022
Preventing Surgery-Induced NK Cell Dysfunction Using Anti-TGF-β Immunotherapeutics.
International journal of molecular sciences
Market M,Tennakoon G,Scaffidi M,Cook DP,Angka L,Ng J,Tanese de Souza C,Kennedy MA,Vanderhyden BC,Auer RC
Published figure using IFN gamma monoclonal antibody (Product # 17-7319-41) in Flow Cytometry
Wed Nov 23 00:00:00 UTC 2022
LINC01123 promotes immune escape by sponging miR-214-3p to regulate B7-H3 in head and neck squamous-cell carcinoma.
Cell death & disease
Li H,Yang Z,Yang X,Zhang F,Wang J,Wu Z,Wanyan C,Meng Q,Gao W,Yang X,Wei J
Published figure using IFN gamma monoclonal antibody (Product # 17-7319-41) in Flow Cytometry
Thu Feb 03 00:00:00 UTC 2022
An Fc-Competent Anti-Human TIGIT Blocking Antibody Ociperlimab (BGB-A1217) Elicits Strong Immune Responses and Potent Anti-Tumor Efficacy in Pre-Clinical Models.
Frontiers in immunology
Chen X,Xue L,Ding X,Zhang J,Jiang L,Liu S,Hou H,Jiang B,Cheng L,Zhu Q,Zhang L,Zhou X,Ma J,Liu Q,Li Y,Ren Z,Jiang B,Song X,Song J,Jin W,Wei M,Shen Z,Liu X,Wang L,Li K,Zhang T
Published figure using IFN gamma monoclonal antibody (Product # 17-7319-41) in Flow Cytometry
Mon May 02 00:00:00 UTC 2022
NRF2/Itaconate Axis Regulates Metabolism and Inflammatory Properties of T Cells in Children with JIA.
Antioxidants (Basel, Switzerland)
Rajendiran A,Subramanyam SH,Klemm P,Jankowski V,van Loosdregt J,Vastert B,Vollbach K,Wagner N,Tenbrock K,Ohl K
Published figure using IFN gamma monoclonal antibody (Product # 17-7319-41) in Flow Cytometry
Thu Dec 08 00:00:00 UTC 2022
A vaccine targeting mutant IDH1 in newly diagnosed glioma.
Nature
Platten M,Bunse L,Wick A,Bunse T,Le Cornet L,Harting I,Sahm F,Sanghvi K,Tan CL,Poschke I,Green E,Justesen S,Behrens GA,Breckwoldt MO,Freitag A,Rother LM,Schmitt A,Schnell O,Hense J,Misch M,Krex D,Stevanovic S,Tabatabai G,Steinbach JP,Bendszus M,von Deimling A,Schmitt M,Wick W
Published figure using IFN gamma monoclonal antibody (Product # 17-7319-41) in Flow Cytometry
Thu Apr 01 00:00:00 UTC 2021
Cancer-secreted exosomal miR-1468-5p promotes tumor immune escape via the immunosuppressive reprogramming of lymphatic vessels.
Molecular therapy : the journal of the American Society of Gene Therapy
Zhou C,Wei W,Ma J,Yang Y,Liang L,Zhang Y,Wang Z,Chen X,Huang L,Wang W,Wu S
Published figure using IFN gamma monoclonal antibody (Product # 17-7319-41) in Flow Cytometry
Wed Apr 07 00:00:00 UTC 2021
Exosome-derived miR-142-5p remodels lymphatic vessels and induces IDO to promote immune privilege in the tumour microenvironment.
Cell death and differentiation
Zhou C,Zhang Y,Yan R,Huang L,Mellor AL,Yang Y,Chen X,Wei W,Wu X,Yu L,Liang L,Zhang D,Wu S,Wang W
Published figure using IFN gamma monoclonal antibody (Product # 17-7319-41) in Flow Cytometry
Mon Feb 01 00:00:00 UTC 2021
Ageing promotes early T follicular helper cell differentiation by modulating expression of RBPJ.
Aging cell
Webb LMC,Fra-Bido S,Innocentin S,Matheson LS,Attaf N,Bignon A,Novarino J,Fazilleau N,Linterman MA
Published figure using IFN gamma monoclonal antibody (Product # 17-7319-41) in Flow Cytometry
Fri Jan 01 00:00:00 UTC 2021
Suppression of 4.1R enhances the potency of NKG2D-CAR T cells against pancreatic carcinoma via activating ERK signaling pathway.
Oncogenesis
Gao Y,Lin H,Guo D,Cheng S,Zhou Y,Zhang L,Yao J,Farooq MA,Ajmal I,Duan Y,He C,Tao L,Wu S,Liu M,Jiang W
Published figure using IFN gamma monoclonal antibody (Product # 17-7319-41) in Flow Cytometry
Tue Sep 21 00:00:00 UTC 2021
Dysregulated Peripheral Invariant Natural Killer T Cells in Plaque Psoriasis Patients.
Frontiers in cell and developmental biology
Hu Y,Chen Y,Chen Z,Zhang X,Guo C,Yu Z,Xu P,Sun L,Zhou X,Gong Y,Yu Q,Shi Y
Published figure using IFN gamma monoclonal antibody (Product # 17-7319-41) in Flow Cytometry
Wed Feb 23 00:00:00 UTC 2022
Identification of CTL Epitopes on Efflux Pumps of the ATP-Binding Cassette and the Major Facilitator Superfamily of Mycobacterium tuberculosis.
Journal of immunology research
Lin Y,Dong Y,Gao Y,Shi R,Li Y,Zhou X,Liu W,Li G,Qi Y,Wu Y
Published figure using IFN gamma monoclonal antibody (Product # 17-7319-41) in Flow Cytometry
Mon Sep 20 00:00:00 UTC 2021
Early loss of T lymphocyte 4-1BB receptor expression is associated with higher short-term mortality in alcoholic hepatitis.
PloS one
Eriksen LL,Nielsen MA,Laursen TL,Deleuran B,Vilstrup H,Støy S
Published figure using IFN gamma monoclonal antibody (Product # 17-7319-41) in Flow Cytometry
Mon Nov 29 00:00:00 UTC 2021
TIGIT+ TIM-3+ NK cells are correlated with NK cell exhaustion and disease progression in patients with hepatitis B virus‑related hepatocellular carcinoma.
Oncoimmunology
Yu L,Liu X,Wang X,Yan F,Wang P,Jiang Y,Du J,Yang Z
Published figure using IFN gamma monoclonal antibody (Product # 17-7319-41) in Flow Cytometry
Mon Aug 02 00:00:00 UTC 2021
A protocol for isolation of primary human immune cells from the liver and mesenteric white adipose tissue biopsies.
STAR protocols
Moreno-Fernandez ME,Damen MSMA,Divanovic S
Published figure using IFN gamma monoclonal antibody (Product # 17-7319-41) in Flow Cytometry
Fri Dec 17 00:00:00 UTC 2021
Suppression of mucosal Th17 memory responses by acellular pertussis vaccines enhances nasal Bordetella pertussis carriage.
NPJ vaccines
Dubois V,Chatagnon J,Thiriard A,Bauderlique-Le Roy H,Debrie AS,Coutte L,Locht C
Published figure using IFN gamma monoclonal antibody (Product # 17-7319-41) in Flow Cytometry
Fri Jan 08 00:00:00 UTC 2021
Niraparib exhibits a synergistic anti-tumor effect with PD-L1 blockade by inducing an immune response in ovarian cancer.
Journal of translational medicine
Meng J,Peng J,Feng J,Maurer J,Li X,Li Y,Yao S,Chu R,Pan X,Li J,Zhang T,Liu L,Zhang Q,Yuan Z,Bu H,Song K,Kong B
Published figure using IFN gamma monoclonal antibody (Product # 17-7319-41) in Flow Cytometry
Thu Oct 07 00:00:00 UTC 2021
Lasting antibody and T cell responses to SARS-CoV-2 in COVID-19 patients three months after infection.
Nature communications
Jiang XL,Wang GL,Zhao XN,Yan FH,Yao L,Kou ZQ,Ji SX,Zhang XL,Li CB,Duan LJ,Li Y,Zhang YW,Duan Q,Wang TC,Li ET,Wei X,Wang QY,Wang XF,Sun WY,Gao YW,Kang DM,Zhang JY,Ma MJ
Published figure using IFN gamma monoclonal antibody (Product # 17-7319-41) in Flow Cytometry
Tue Feb 09 00:00:00 UTC 2021
Identification of HLA-A2-Restricted Mutant Epitopes from Neoantigens of Esophageal Squamous Cell Carcinoma.
Vaccines
Wang Z,Ran L,Chen C,Shi R,Dong Y,Li Y,Zhou X,Qi Y,Zhu P,Gao Y,Wu Y
Published figure using IFN gamma monoclonal antibody (Product # 17-7319-41) in Flow Cytometry
Fri Oct 01 00:00:00 UTC 2021
NKD2 mediates stimulation-dependent ORAI1 trafficking to augment Ca2+ entry in T cells.
Cell reports
Wu B,Woo JS,Vila P,Jew M,Leung J,Sun Z,Srikanth S,Gwack Y
Published figure using IFN gamma monoclonal antibody (Product # 17-7319-41) in Flow Cytometry
Tue Aug 24 00:00:00 UTC 2021
Multipeptide stimulated PBMCs generate TEM/TCM for adoptive cell therapy in multiple myeloma.
Oncotarget
Vardam-Kaur T,Pathangey LB,McCormick DJ,Bergsagel PL,Cohen PA,Gendler SJ
Published figure using IFN gamma monoclonal antibody (Product # 17-7319-41) in Flow Cytometry
Tue Sep 28 00:00:00 UTC 2021
YY1 regulation by miR-124-3p promotes Th17 cell pathogenicity through interaction with T-bet in rheumatoid arthritis.
JCI insight
Lin J,Tang J,Lin J,He Y,Yu Z,Jiang R,Yang B,Ou Q
Published figure using IFN gamma monoclonal antibody (Product # 17-7319-41) in Flow Cytometry
Mon Nov 22 00:00:00 UTC 2021
Rejuvenating Effector/Exhausted CAR T Cells to Stem Cell Memory-Like CAR T Cells By Resting Them in the Presence of CXCL12 and the NOTCH Ligand.
Cancer research communications
Ando M,Kondo T,Tomisato W,Ito M,Shichino S,Srirat T,Mise-Omata S,Nakagawara K,Yoshimura A
Published figure using IFN gamma monoclonal antibody (Product # 17-7319-41) in Flow Cytometry
Fri Oct 01 00:00:00 UTC 2021
Inhibition of Cholesterol Esterification Enzyme Enhances the Potency of Human Chimeric Antigen Receptor T Cells against Pancreatic Carcinoma.
Molecular therapy oncolytics
Zhao L,Liu Y,Zhao F,Jin Y,Feng J,Geng R,Sun J,Kang L,Yu L,Wei Y
Published figure using IFN gamma monoclonal antibody (Product # 17-7319-41) in Flow Cytometry
Fri Mar 27 00:00:00 UTC 2020
DEK-targeting aptamer DTA-64 attenuates bronchial EMT-mediated airway remodelling by suppressing TGF-β1/Smad, MAPK and PI3K signalling pathway in asthma.
Journal of cellular and molecular medicine
Song Y,Wang Z,Jiang J,Piao Y,Li L,Xu C,Piao H,Li L,Yan G
17-7319 was used in Flow cytometry/Cell sorting to study DEK-targeting aptamer in EMT-mediated airway remodeling in asthma.
Tue Dec 01 00:00:00 UTC 2020
DEK-targeting aptamer DTA-64 attenuates bronchial EMT-mediated airway remodelling by suppressing TGF-β1/Smad, MAPK and PI3K signalling pathway in asthma.
Journal of cellular and molecular medicine
Song Y,Wang Z,Jiang J,Piao Y,Li L,Xu C,Piao H,Li L,Yan G
17-7319 was used in Flow cytometry/Cell sorting to study DEK-targeting aptamer in EMT-mediated airway remodeling in asthma.
Tue Dec 01 00:00:00 UTC 2020
TIRC7 inhibits Th1 cells by upregulating the expression of CTLA‑4 and STAT3 in mice with acute graft‑versus‑host disease.
Oncology reports
Zhu F,Qiu T,Zhu S,Zhao K,Chen C,Qiao J,Pan B,Yan Z,Chen W,Liu Q,Wu Q,Cao J,Sang W,Zeng L,Sun H,Li Z,Xu K
Published figure using IFN gamma monoclonal antibody (Product # 17-7319-41) in Flow Cytometry
Wed Jul 01 00:00:00 UTC 2020
TIRC7 inhibits Th1 cells by upregulating the expression of CTLA‑4 and STAT3 in mice with acute graft‑versus‑host disease.
Oncology reports
Zhu F,Qiu T,Zhu S,Zhao K,Chen C,Qiao J,Pan B,Yan Z,Chen W,Liu Q,Wu Q,Cao J,Sang W,Zeng L,Sun H,Li Z,Xu K
Published figure using IFN gamma monoclonal antibody (Product # 17-7319-41) in Flow Cytometry
Wed Jul 01 00:00:00 UTC 2020
Immunoprophylactic and immunotherapeutic control of hormone receptor-positive breast cancer.
Nature communications
Buqué A,Bloy N,Perez-Lanzón M,Iribarren K,Humeau J,Pol JG,Levesque S,Mondragon L,Yamazaki T,Sato A,Aranda F,Durand S,Boissonnas A,Fucikova J,Senovilla L,Enot D,Hensler M,Kremer M,Stoll G,Hu Y,Massa C,Formenti SC,Seliger B,Elemento O,Spisek R,André F,Zitvogel L,Delaloge S,Kroemer G,Galluzzi L
Published figure using IFN gamma monoclonal antibody (Product # 17-7319-41) in Flow Cytometry
Thu Jul 30 00:00:00 UTC 2020
Single-cell transcriptomics identifies an effectorness gradient shaping the response of CD4+ T cells to cytokines.
Nature communications
Cano-Gamez E,Soskic B,Roumeliotis TI,So E,Smyth DJ,Baldrighi M,Willé D,Nakic N,Esparza-Gordillo J,Larminie CGC,Bronson PG,Tough DF,Rowan WC,Choudhary JS,Trynka G
Published figure using IFN gamma monoclonal antibody (Product # 17-7319-41) in Flow Cytometry
Tue Apr 14 00:00:00 UTC 2020
Ovarian granulosa cell tumor characterization identifies FOXL2 as an immunotherapeutic target.
JCI insight
Pierini S,Tanyi JL,Simpkins F,George E,Uribe-Herranz M,Drapkin R,Burger R,Morgan MA,Facciabene A
Published figure using IFN gamma monoclonal antibody (Product # 17-7319-41) in Flow Cytometry
Thu Aug 20 00:00:00 UTC 2020
A Method of Assessment of Human Natural Killer Cell Phenotype and Function in Whole Blood.
Frontiers in immunology
Market M,Tennakoon G,Ng J,Scaffidi M,de Souza CT,Kennedy MA,Auer RC
Published figure using IFN gamma monoclonal antibody (Product # 17-7319-41) in Flow Cytometry
Mon Mar 29 00:00:00 UTC 2021
Human Tumor-Infiltrating MAIT Cells Display Hallmarks of Bacterial Antigen Recognition in Colorectal Cancer.
Cell reports. Medicine
Li S,Simoni Y,Becht E,Loh CY,Li N,Lachance D,Koo SL,Lim TP,Tan EKW,Mathew R,Nguyen A,Golovato J,Berkson JD,Prlic M,Lee B,Minot SS,Nagarajan N,Dey N,Tan DSW,Tan IB,Newell EW
Published figure using IFN gamma monoclonal antibody (Product # 17-7319-41) in Flow Cytometry
Tue Jun 23 00:00:00 UTC 2020
NPM1 upregulates the transcription of PD-L1 and suppresses T cell activity in triple-negative breast cancer.
Nature communications
Qin G,Wang X,Ye S,Li Y,Chen M,Wang S,Qin T,Zhang C,Li Y,Long Q,Hu H,Shi D,Li J,Zhang K,Zhai Q,Tang Y,Kang T,Lan P,Xie F,Lu J,Deng W
Published figure using IFN gamma monoclonal antibody (Product # 17-7319-41) in Flow Cytometry
Fri Apr 03 00:00:00 UTC 2020
NPM1 upregulates the transcription of PD-L1 and suppresses T cell activity in triple-negative breast cancer.
Nature communications
Qin G,Wang X,Ye S,Li Y,Chen M,Wang S,Qin T,Zhang C,Li Y,Long Q,Hu H,Shi D,Li J,Zhang K,Zhai Q,Tang Y,Kang T,Lan P,Xie F,Lu J,Deng W
Published figure using IFN gamma monoclonal antibody (Product # 17-7319-41) in Flow Cytometry
Fri Apr 03 00:00:00 UTC 2020
CAIX Regulates GBM Motility and TAM Adhesion and Polarization through EGFR/STAT3 under Hypoxic Conditions.
International journal of molecular sciences
Huang BR,Liu YS,Lai SW,Lin HJ,Shen CK,Yang LY,Lu DY
Published figure using IFN gamma monoclonal antibody (Product # 17-7319-41) in Flow Cytometry
Fri Aug 14 00:00:00 UTC 2020
Duration of Cellular and Humoral Responses after Quadrivalent Human Papillomavirus Vaccination in Healthy Female Adults with or without Prior Type 16 and/or 18 Exposure.
Vaccines
Lai L,Ault K,Rouphael N,Beck A,Domjahn B,Xu Y,Anderson EJ,Cheng A,Nakamura A,Hoagland RJ,Kelley C,Edupuganti S,Mask K,Nesin M,Unger ER,Panicker G,David H,Mulligan MJ
Published figure using IFN gamma monoclonal antibody (Product # 17-7319-41) in Flow Cytometry
Tue Jun 30 00:00:00 UTC 2020
Th2 Biased Immunity With Altered B Cell Profiles in Circulation of Patients With Sporotrichosis Caused by Sporothrix globosa.
Frontiers in immunology
Zu J,Yao L,Song Y,Cui Y,Guan M,Chen R,Zhen Y,Li S
Published figure using IFN gamma monoclonal antibody (Product # 17-7319-41) in Flow Cytometry
Mon May 31 00:00:00 UTC 2021
Mouse Double Minute 2 Homolog-Mediated Ubiquitination Facilitates Forkhead Box P3 Stability and Positively Modulates Human Regulatory T Cell Function.
Frontiers in immunology
Wang A,Yang M,Liang R,Zhu F,Zhu F,Liu X,Han Y,Lin R,Wang X,Li D,Li H,Yuan X,Zhao H,Li B
17-7319 was used in Flow cytometry/Cell sorting to investigate how mouse double minute 2 homolog effects the function of regulatory T cells.
Mon Apr 05 00:00:00 UTC 2021
Dynamic single-cell phenotyping of immune cells using the microfluidic platform DropMap.
Nature protocols
Bounab Y,Eyer K,Dixneuf S,Rybczynska M,Chauvel C,Mistretta M,Tran T,Aymerich N,Chenon G,Llitjos JF,Venet F,Monneret G,Gillespie IA,Cortez P,Moucadel V,Pachot A,Troesch A,Leissner P,Textoris J,Bibette J,Guyard C,Baudry J,Griffiths AD,Védrine C
17-7319-41 was used in Flow Cytometry to develop a microfluidic platform, DropMap, for the phenotyping of different immune cell types.
Tue Sep 01 00:00:00 UTC 2020
Chronic Inflammation Permanently Reshapes Tissue-Resident Immunity in Celiac Disease.
Cell
Mayassi T,Ladell K,Gudjonson H,McLaren JE,Shaw DG,Tran MT,Rokicka JJ,Lawrence I,Grenier JC,van Unen V,Ciszewski C,Dimaano M,Sayegh HE,Kumar V,Wijmenga C,Green PHR,Gokhale R,Jericho H,Semrad CE,Guandalini S,Dinner AR,Kupfer SS,Reid HH,Barreiro LB,Rossjohn J,Price DA,Jabri B
17-7319-41 was used in Flow Cytometry to determine the role of tissue-resident lymphocytes in chronic inflammation.
Thu Feb 21 00:00:00 UTC 2019
Subsets of ILC3-ILC1-like cells generate a diversity spectrum of innate lymphoid cells in human mucosal tissues.
Nature immunology
Cella M,Gamini R,Sécca C,Collins PL,Zhao S,Peng V,Robinette ML,Schettini J,Zaitsev K,Gordon W,Bando JK,Yomogida K,Cortez V,Fronick C,Fulton R,Lin LL,Gilfillan S,Flavell RA,Shan L,Artyomov MN,Bowman M,Oltz EM,Jelinsky SA,Colonna M
17-7319 was used in Flow cytometry/Cell sorting to demonstrate that innate lymphoid cells (ILCs) undergo conversion from ILC3s into ILC1-like cells in human tissues in vivo, requiring tissue factors and Aiolos in the process.
Thu Aug 01 00:00:00 UTC 2019
Engineered triple inhibitory receptor resistance improves anti-tumor CAR-T cell performance via CD56.
Nature communications
Zou F,Lu L,Liu J,Xia B,Zhang W,Hu Q,Liu W,Zhang Y,Lin Y,Jing S,Huang M,Huang B,Liu B,Zhang H
Published figure using IFN gamma monoclonal antibody (Product # 17-7319-41) in Flow Cytometry
Wed Sep 11 00:00:00 UTC 2019
CD4 T cell loss and Th2 and Th17 bias are associated with the severity of severe fever with thrombocytopenia syndrome (SFTS).
Clinical immunology (Orlando, Fla.)
Li MM,Zhang WJ,Weng XF,Li MY,Liu J,Xiong Y,Xiong SE,Zou CC,Wang H,Lu MJ,Yang DL,Peng C,Zheng X
Published figure using IFN gamma monoclonal antibody (Product # 17-7319-41) in Flow Cytometry
Mon Oct 01 00:00:00 UTC 2018
RKIP mediates autoimmune inflammation by positively regulating IL-17R signaling.
EMBO reports
Lin W,Wang N,Zhou K,Su F,Jiang Y,Shou J,Liu H,Ma C,Qian Y,Wang K,Wang X
Published figure using IFN gamma monoclonal antibody (Product # 17-7319-41) in Flow Cytometry
Fri Jun 01 00:00:00 UTC 2018
CXCL4 is a novel inducer of human Th17 cells and correlates with IL-17 and IL-22 in psoriatic arthritis.
European journal of immunology
Affandi AJ,Silva-Cardoso SC,Garcia S,Leijten EFA,van Kempen TS,Marut W,van Roon JAG,Radstake TRDJ
Published figure using IFN gamma monoclonal antibody (Product # 17-7319-41) in Flow Cytometry
Thu Mar 01 00:00:00 UTC 2018
Accumulation of T-helper 22 cells, interleukin-22 and myeloid-derived suppressor cells promotes gastric cancer progression in elderly patients.
Oncology letters
Chen X,Wang Y,Wang J,Wen J,Jia X,Wang X,Zhang H
Published figure using IFN gamma monoclonal antibody (Product # 17-7319-41) in Flow Cytometry
Sun Jul 01 00:00:00 UTC 2018
Chloroquine modulates antitumor immune response by resetting tumor-associated macrophages toward M1 phenotype.
Nature communications
Chen D,Xie J,Fiskesund R,Dong W,Liang X,Lv J,Jin X,Liu J,Mo S,Zhang T,Cheng F,Zhou Y,Zhang H,Tang K,Ma J,Liu Y,Huang B
Published figure using IFN gamma monoclonal antibody (Product # 17-7319-41) in Flow Cytometry
Wed Feb 28 00:00:00 UTC 2018
Human in vivo-generated monocyte-derived dendritic cells and macrophages cross-present antigens through a vacuolar pathway.
Nature communications
Tang-Huau TL,Gueguen P,Goudot C,Durand M,Bohec M,Baulande S,Pasquier B,Amigorena S,Segura E
Published figure using IFN gamma monoclonal antibody (Product # 17-7319-41) in Flow Cytometry
Mon Jul 02 00:00:00 UTC 2018
Impact of Enhancer of Zeste Homolog 2 on T Helper Cell-Mediated Allergic Rhinitis.
Frontiers in immunology
Hou TY,Chen MR,Chou YC,Kan PC,Tsai YT,Cha TL
Published figure using IFN gamma monoclonal antibody (Product # 17-7319-41) in Flow Cytometry
Wed Nov 20 00:00:00 UTC 2019
Ebola virus glycoprotein directly triggers T lymphocyte death despite of the lack of infection.
PLoS pathogens
Iampietro M,Younan P,Nishida A,Dutta M,Lubaki NM,Santos RI,Koup RA,Katze MG,Bukreyev A
Published figure using IFN gamma monoclonal antibody (Product # 17-7319-41) in Flow Cytometry
Mon May 01 00:00:00 UTC 2017
Dominant immunosuppression of dendritic cell function by prostate-cancer-derived exosomes.
Journal of extracellular vesicles
Salimu J,Webber J,Gurney M,Al-Taei S,Clayton A,Tabi Z
Published figure using IFN gamma monoclonal antibody (Product # 17-7319-41) in Flow Cytometry
Wed Sep 30 00:00:00 UTC 2020
Single-cell profiling reveals GPCR heterogeneity and functional patterning during neuroinflammation.
JCI insight
Tischner D,Grimm M,Kaur H,Staudenraus D,Carvalho J,Looso M,Günther S,Wanke F,Moos S,Siller N,Breuer J,Schwab N,Zipp F,Waisman A,Kurschus FC,Offermanns S,Wettschureck N
Published figure using IFN gamma monoclonal antibody (Product # 17-7319-41) in Flow Cytometry
Thu Aug 03 00:00:00 UTC 2017
Surrogate in vitro activation of innate immunity synergizes with interleukin-7 to unleash rapid antigen-driven outgrowth of CD4+ and CD8+ human peripheral blood T-cells naturally recognizing MUC1, HER2/neu and other tumor-associated antigens.
Oncotarget
Pathangey LB,McCurry DB,Gendler SJ,Dominguez AL,Gorman JE,Pathangey G,Mihalik LA,Dang Y,Disis ML,Cohen PA
Published figure using IFN gamma monoclonal antibody (Product # 17-7319-41) in Flow Cytometry
Tue Feb 14 00:00:00 UTC 2017
NKG2C(+)CD57(+) Natural Killer Cell Expansion Parallels Cytomegalovirus-Specific CD8(+) T Cell Evolution towards Senescence.
Journal of immunology research
Heath J,Newhook N,Comeau E,Gallant M,Fudge N,Grant M
17-7319 was used in Flow cytometry/Cell sorting to explore the relationship between NKG2C(+)CD57(+) natural killer cells and the adaptive immune response to human cytomegalovirus.
Thu Mar 16 00:00:00 UTC 2017
CD28 family of receptors on T cells in chronic HBV infection: Expression characteristics, clinical significance and correlations with PD-1 blockade.
Molecular medicine reports
Tang ZS,Hao YH,Zhang EJ,Xu CL,Zhou Y,Zheng X,Yang DL
Published figure using IFN gamma monoclonal antibody (Product # 17-7319-41) in Flow Cytometry
Mon Aug 01 00:00:00 UTC 2016
Heterologous Immunity between Adenoviruses and Hepatitis C Virus: A New Paradigm in HCV Immunity and Vaccines.
PloS one
Singh S,Vedi S,Samrat SK,Li W,Kumar R,Agrawal B
Published figure using IFN gamma monoclonal antibody (Product # 17-7319-41) in Flow Cytometry
Tue Jul 19 00:00:00 UTC 2016
Peritoneal carcinomatosis of colorectal cancer is characterized by structural and functional reorganization of the tumor microenvironment inducing senescence and proliferation arrest in cancer cells.
Oncoimmunology
Seebauer CT,Brunner S,Glockzin G,Piso P,Ruemmele P,Schlitt HJ,Geissler EK,Fichtner-Feigl S,Kesselring R
Published figure using IFN gamma monoclonal antibody (Product # 17-7319-41) in Flow Cytometry
Tue Jan 12 00:00:00 UTC 2021
FOXP3+ Regulatory T Cells in Hepatic Fibrosis and Splenomegaly Caused by Schistosoma japonicum: The Spleen May Be a Major Source of Tregs in Subjects with Splenomegaly.
PLoS neglected tropical diseases
Romano A,Hou X,Sertorio M,Dessein H,Cabantous S,Oliveira P,Li J,Oyegue S,Arnaud V,Luo X,Daujat-Chavanieu M,Mariani O,Sastre X,Dombey AM,He H,Li Y,Dessein A
17-7319 was used in Flow cytometry/Cell sorting to investigate whether FOXP3+ Tregs play a role in human schistosomiasis.
Fri Jan 01 00:00:00 UTC 2016
A SnoRNA-derived piRNA interacts with human interleukin-4 pre-mRNA and induces its decay in nuclear exosomes.
Nucleic acids research
Zhong F,Zhou N,Wu K,Guo Y,Tan W,Zhang H,Zhang X,Geng G,Pan T,Luo H,Zhang Y,Xu Z,Liu J,Liu B,Gao W,Liu C,Ren L,Li J,Zhou J,Zhang H
Published figure using IFN gamma monoclonal antibody (Product # 17-7319-41) in Flow Cytometry
Wed Dec 02 00:00:00 UTC 2015
Protective genotypes in HIV infection reflect superior function of KIR3DS1+ over KIR3DL1+ CD8+ T cells.
Immunology and cell biology
Zipperlen K,Gallant M,Stapleton S,Heath J,Barrett L,Grant M
17-7319 was used in Flow cytometry/Cell sorting to investigate how HLA/KIR genotypic combinations relate to the phenotype and function of CD8(+) T cells in individuals with HIV infection.
Thu Jan 01 00:00:00 UTC 2015
Type I IFNs and IL-18 regulate the antiviral response of primary human γδ T cells against dendritic cells infected with Dengue virus.
Journal of immunology (Baltimore, Md. : 1950)
Tsai CY,Liong KH,Gunalan MG,Li N,Lim DS,Fisher DA,MacAry PA,Leo YS,Wong SC,Puan KJ,Wong SB
17-7319 was used in Flow cytometry/Cell sorting to investigate the markers of cellular innate immunity against dengue virus, specifically how type I IFNs and IL-18 regulate activation, proliferation and degranulation of T cells against dendritic cells in human patients with dengue virus.
Wed Apr 15 00:00:00 UTC 2015
Expression of GM-CSF in T Cells Is Increased in Multiple Sclerosis and Suppressed by IFN-β Therapy.
Journal of immunology (Baltimore, Md. : 1950)
Rasouli J,Ciric B,Imitola J,Gonnella P,Hwang D,Mahajan K,Mari ER,Safavi F,Leist TP,Zhang GX,Rostami A
17-7319 was used in Flow cytometry/Cell sorting to characterise GM-CSF production by T cells of MS patients and to determine the effect of IFN-β therapy on its production.
Mon Jun 01 00:00:00 UTC 2015
Induced expression of FcγRIIIa (CD16a) on CD4+ T cells triggers generation of IFN-γhigh subset.
The Journal of biological chemistry
Chauhan AK,Chen C,Moore TL,DiPaolo RJ
17-7319 was used in Flow cytometry/Cell sorting to describe for the first time a role for FcγRIIIa in CD4+ T-cell differentiation.
Fri Feb 20 00:00:00 UTC 2015
Induced expression of FcγRIIIa (CD16a) on CD4+ T cells triggers generation of IFN-γhigh subset.
The Journal of biological chemistry
Chauhan AK,Chen C,Moore TL,DiPaolo RJ
17-7319 was used in Flow cytometry/Cell sorting to describe for the first time a role for FcγRIIIa in CD4+ T-cell differentiation.
Fri Feb 20 00:00:00 UTC 2015
Multifactorial T-cell hypofunction that is reversible can limit the efficacy of chimeric antigen receptor-transduced human T cells in solid tumors.
Clinical cancer research : an official journal of the American Association for Cancer Research
Moon EK,Wang LC,Dolfi DV,Wilson CB,Ranganathan R,Sun J,Kapoor V,Scholler J,Puré E,Milone MC,June CH,Riley JL,Wherry EJ,Albelda SM
Published figure using IFN gamma monoclonal antibody (Product # 17-7319-41) in Flow Cytometry
Fri Aug 15 00:00:00 UTC 2014
PD-1(+) CD8(+) T cells are exhausted in tumours and functional in draining lymph nodes of colorectal cancer patients.
British journal of cancer
Wu X,Zhang H,Xing Q,Cui J,Li J,Li Y,Tan Y,Wang S
17-7319 was used in Immunohistochemistry to evaluate the role of PD-1-PD-L1 pathway in CD8(+) T-cell functions in tumour-draining lymph nodes (TDLNs) and tumours of CRC patients.
Tue Sep 23 00:00:00 UTC 2014
PD-1(+) CD8(+) T cells are exhausted in tumours and functional in draining lymph nodes of colorectal cancer patients.
British journal of cancer
Wu X,Zhang H,Xing Q,Cui J,Li J,Li Y,Tan Y,Wang S
17-7319 was used in Immunohistochemistry to evaluate the role of PD-1-PD-L1 pathway in CD8(+) T-cell functions in tumour-draining lymph nodes (TDLNs) and tumours of CRC patients.
Tue Sep 23 00:00:00 UTC 2014
Failure to upregulate cell surface PD-1 is associated with dysregulated stimulation of T cells by TGN1412-like CD28 superagonist.
mAbs
Thaventhiran T,Alhumeed N,Yeang HX,Sethu S,Downey JS,Alghanem AF,Olayanju A,Smith EL,Cross MJ,Webb SD,Williams DP,Bristow A,Ball C,Stebbings R,Sathish JG
17-7319 was used in Flow cytometry/Cell sorting to understand the underlying molecular mechanisms of T-cell activation in response to TGN1412-like CD28 superagonist, showing that failure to upregulate the inhibitory PD-1 cell surface receptor is part of this mechanism.
Mon Aug 24 00:00:00 UTC 2015
Failure to upregulate cell surface PD-1 is associated with dysregulated stimulation of T cells by TGN1412-like CD28 superagonist.
mAbs
Thaventhiran T,Alhumeed N,Yeang HX,Sethu S,Downey JS,Alghanem AF,Olayanju A,Smith EL,Cross MJ,Webb SD,Williams DP,Bristow A,Ball C,Stebbings R,Sathish JG
17-7319 was used in Flow cytometry/Cell sorting to understand the underlying molecular mechanisms of T-cell activation in response to TGN1412-like CD28 superagonist, showing that failure to upregulate the inhibitory PD-1 cell surface receptor is part of this mechanism.
Mon Aug 24 00:00:00 UTC 2015
Human γδ thymocytes are functionally immature and differentiate into cytotoxic type 1 effector T cells upon IL-2/IL-15 signaling.
Journal of immunology (Baltimore, Md. : 1950)
Ribot JC,Ribeiro ST,Correia DV,Sousa AE,Silva-Santos B
17-7319 was used in Flow cytometry/Cell sorting to study how ex vivo-isolated γδ thymocytes produced negligible IFN-γ and lacked cytolytic activity against leukemia cells.
Sat Mar 01 00:00:00 UTC 2014
The combination of type I IFN, TNF-α, and cell surface receptor engagement with dendritic cells enables NK cells to overcome immune evasion by dengue virus.
Journal of immunology (Baltimore, Md. : 1950)
Lim DS,Yawata N,Selva KJ,Li N,Tsai CY,Yeong LH,Liong KH,Ooi EE,Chong MK,Ng ML,Leo YS,Yawata M,Wong SB
17-7319 was used in Flow cytometry/Cell sorting to investigate the response of NK cells against dengue-infected cells, showing that a combination of type I IFN, TNF-α, and cell surface receptor engagement with dendritic cells is required.
Sat Nov 15 00:00:00 UTC 2014
Tumor-derived exosomes promote tumor progression and T-cell dysfunction through the regulation of enriched exosomal microRNAs in human nasopharyngeal carcinoma.
Oncotarget
Ye SB,Li ZL,Luo DH,Huang BJ,Chen YS,Zhang XS,Cui J,Zeng YX,Li J
17-7319 was used in Flow cytometry/Cell sorting to show that tumour-derived exosomes promote tumour progression and T-cell dysfunction through regulation of exosomal microRNAs in human carcinomas.
Wed Jul 30 00:00:00 UTC 2014
BET bromodomain inhibition suppresses TH17-mediated pathology.
The Journal of experimental medicine
Mele DA,Salmeron A,Ghosh S,Huang HR,Bryant BM,Lora JM
17-7319 was used in Flow cytometry/Cell sorting to identify BET proteins as a fundamental link between chromatin signalling and T(H)17 biology.
Mon Oct 21 00:00:00 UTC 2013
Memory T cells in latent Mycobacterium tuberculosis infection are directed against three antigenic islands and largely contained in a CXCR3+CCR6+ Th1 subset.
PLoS pathogens
Lindestam Arlehamn CS,Gerasimova A,Mele F,Henderson R,Swann J,Greenbaum JA,Kim Y,Sidney J,James EA,Taplitz R,McKinney DM,Kwok WW,Grey H,Sallusto F,Peters B,Sette A
17-7319 was used in Flow cytometry/Cell sorting to further define the immunological footprint of Mycobacterium tuberculosis CD4 T cell recognition.
Tue Jan 01 00:00:00 UTC 2013
Elevated frequencies of circulating Th22 cell in addition to Th17 cell and Th17/Th1 cell in patients with acute coronary syndrome.
PloS one
Zhang L,Wang T,Wang XQ,Du RZ,Zhang KN,Liu XG,Ma DX,Yu S,Su GH,Li ZH,Guan YQ,Du NL
Published figure using IFN gamma monoclonal antibody (Product # 17-7319-41) in Flow Cytometry
Mon Aug 18 00:00:00 UTC 2014
CD25 blockade depletes and selectively reprograms regulatory T cells in concert with immunotherapy in cancer patients.
Science translational medicine
Rech AJ,Mick R,Martin S,Recio A,Aqui NA,Powell DJ,Colligon TA,Trosko JA,Leinbach LI,Pletcher CH,Tweed CK,DeMichele A,Fox KR,Domchek SM,Riley JL,Vonderheide RH
17-7319 was used in Flow cytometry/Cell sorting to conclude that CD25 blockade depletes and selectively reprograms T(regs) in concert with active immune therapy in cancer patients.
Wed May 16 00:00:00 UTC 2012
Peripherally circulating CD4⁺ FOXP3⁺ CXCR3⁺ T regulatory cells correlate with renal allograft function.
Scandinavian journal of immunology
Hoerning A,Köhler S,Jun C,Tebbe B,Fu J,Menke J,Wilde B,Dolff S,Feldkamp T,Briscoe DM,Kribben A,Hoyer PF,Witzke O
17-7319 was used in Flow cytometry/Cell sorting to investigate the relevance of trafficking receptors for Tregs after solid organ transplantation.
Sat Sep 01 00:00:00 UTC 2012
Brefeldin A, but not monensin, enables flow cytometric detection of interleukin-4 within peripheral T cells responding to ex vivo stimulation with Chlamydia trachomatis.
Journal of immunological methods
Vicetti Miguel RD,Maryak SA,Cherpes TL
17-7319 was used in Flow cytometry/Cell sorting to compare the ability of brefeldin A and monensin to concurrently trap 4 cytokines within peripheral blood mononuclear cells.
Wed Oct 31 00:00:00 UTC 2012
Increased frequencies of Th22 cells as well as Th17 cells in the peripheral blood of patients with ankylosing spondylitis and rheumatoid arthritis.
PloS one
Zhang L,Li YG,Li YH,Qi L,Liu XG,Yuan CZ,Hu NW,Ma DX,Li ZF,Yang Q,Li W,Li JM
Published figure using IFN gamma monoclonal antibody (Product # 17-7319-41) in Flow Cytometry
Fri Jul 27 00:00:00 UTC 2012
Role played by CD4+FOXP3+ regulatory T Cells in suppression of host responses to Haemophilus ducreyi during experimental infection of human volunteers.
The Journal of infectious diseases
Li W,Tenner-Racz K,Racz P,Janowicz DM,Fortney KR,Katz BP,Spinola SM
17-7319 was used in Flow cytometry/Cell sorting to propose that the population of CD4(+)CD25(+)CD127(lo/-)FOXP3(+) T(reg) cells are expanded at H. ducreyi-infected sites and may supress the hosts immune response.
Tue Jun 15 00:00:00 UTC 2010
CXCL10 production by human monocytes in response to Leishmania braziliensis infection.
Infection and immunity
Vargas-Inchaustegui DA,Hogg AE,Tulliano G,Llanos-Cuentas A,Arevalo J,Endsley JJ,Soong L
17-7319 was used in Flow cytometry/Cell sorting to investigate whether unbalance production of inflammatory mediators in response to L. braziliensis infection contributes to disease severity, showing that CXCL10 is produced in response to infection.
Fri Jan 01 00:00:00 UTC 2010
Human immunodeficiency virus type I-specific CD8+ T cell subset abnormalities in chronic infection persist through effective antiretroviral therapy.
BMC infectious diseases
Pohling J,Zipperlen K,Hollett NA,Gallant ME,Grant MD
17-7319 was used in Flow cytometry/Cell sorting to evaluate the magnitude, general specificity and character of HIV specific CD8+ T cell responses, showing CD8+ T cell abnormalities in chronic infection.
Tue May 25 00:00:00 UTC 2010
Phenotypic analysis of prostate-infiltrating lymphocytes reveals TH17 and Treg skewing.
Clinical cancer research : an official journal of the American Association for Cancer Research
Sfanos KS,Bruno TC,Maris CH,Xu L,Thoburn CJ,DeMarzo AM,Meeker AK,Isaacs WB,Drake CG
17-7319 was used in Flow cytometry/Cell sorting to reveal a potential role for TH17 and regulatory CD4+ T cells in the development or progression of prostate cancer.
Sun Jun 01 00:00:00 UTC 2008