Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Description: Anti-human IFN-omega (OMG-5) neutralize the antiviral activity of IFN-omega.
In the human genome, the IFN-omega gene family consists of seven members located on chromosome 9. However, only one of these genes is functional giving rise to the IFN-omega protein, whereas the others are non-functional pseudogenes. The IFN-omega gene is not expressed in unstimulated cells. Viral infection results in expression of the gene giving rise to the N-glycosylated protein consisting of 172 or 174 amino acids and an apparent molecular mass of about 25kDa. A single carbohydrate group consists mainly of biantennary complex oligosaccharides with variable amounts of N-acetyl neuraminic acid. In quantitative terms, IFN-omega is a major component of human leukocyte IFN; with a contribution to its total antiviral activity estimated to be in the range of 10 -15 %.
Applications Tested: ELISA.
IFN-omega is a type I interferon that can be induced by virus-infected leukocytes. Members of the type I interferon family, which includes IFN-alpha, IFN-beta, and IFN-omega, signal through the IFNAR-1/IFNAR-2 receptor complex, and exert antiviral and antiproliferative activities. IFN-omega exhibits about 75% sequence homology with IFN-alpha, and contains two conserved disulfide bonds that are necessary for full biological activity.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: IFN o; IFN ω; IFN-omega 1, interferon omega-1; IFNo; IFNomega; IFNW; IFNω; Interferon alpha-II-1; Interferon o; Interferon OMEGA; Interferon omega-1; Interferon ω; interferon, omega 1
Gene Aliases: IFNW1
UniProt ID: (Human) P05000
Entrez Gene ID: (Human) 3467
Molecular Function:
cytokine
interferon superfamily
intercellular signal molecule
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support