Search Thermo Fisher Scientific
- Contáctenos
- Orden Rápida
-
¿No tiene una cuenta? Crear una cuenta
Search Thermo Fisher Scientific
Immunogen sequence: AWVTMPWFQ HYRNVTVGPP ENIEVTPGEG SLIIRFSSPF DIADTSTAFF CYYVHYWEKG GIQQVKGPFR SNSISLDNLK PSRVYCLQVQ AQLLWNKSNI FRVGHLSNIS CYETMADAST ELQQVILISV GTFSLLSVLA GACFFLVLKY RGLIKYWFHT PPSIPLQIEE YLKDPTQPIL EALDKDSSPK DDVWDSVSII S (125-324 aa encoded by BC003624)
This gene (IFNGR2) encodes the non-ligand-binding beta chain of the gamma interferon receptor. Human interferon-gamma receptor is a heterodimer of IFNGR1 and IFNGR2. Defects in IFNGR2 are a cause of mendelian susceptibility to mycobacterial disease (MSMD), also known as familial disseminated atypical mycobacterial infection. MSMD is a genetically heterogeneous disease with autosomal recessive, autosomal dominant or X-linked inheritance.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: AF-1; IFN-gamma receptor 2; IFN-gamma-R-beta; IFN-gamma-R2; Interferon gamma receptor 2; Interferon gamma receptor accessory factor 1; interferon gamma receptor accessory factor-1; interferon gamma receptor beta chain; Interferon gamma receptor beta-chain; Interferon gamma transducer 1
Gene Aliases: AF-1; IFGR2; IFNGR2; IFNGT1; IMD28
UniProt ID: (Human) P38484
Entrez Gene ID: (Human) 3460
Molecular Function: transmembrane signal receptor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support