Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MRPTLLWSLL LLLGVFAAAA AAPPDPLSQL PAPQHPKIRL YNAEQVLSWE PVALSNSTRP VVYQVQFKYT DSKWFTADIM SIGVNCTQIT ATECDFTAAS PSAGFPMDFN VTLRLRAELG ALHSAWVTMP WFQHYRNVTV GPPENIEVTP GEGSLIIRFS SPFDIADTST AFFCYYVHYW EKGGIQQVKG PFRSNSISLD NLKPSRVYCL QVQAQLLWNK SNIFRVGHLS NISCYETMAD ASTELQQVIL ISVGTFSLLS VLAGACFFLV LKYRGLIKYW FHTPPSIPLQ IEEYLKDPTQ PILEALDKDS SPKDDVWDSV SIISFPEKEQ EDVLQTL
This gene (IFNGR2) encodes the non-ligand-binding beta chain of the gamma interferon receptor. Human interferon-gamma receptor is a heterodimer of IFNGR1 and IFNGR2. Defects in IFNGR2 are a cause of mendelian susceptibility to mycobacterial disease (MSMD), also known as familial disseminated atypical mycobacterial infection. MSMD is a genetically heterogeneous disease with autosomal recessive, autosomal dominant or X-linked inheritance.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: AF-1; IFN-gamma receptor 2; IFN-gamma-R-beta; IFN-gamma-R2; Interferon gamma receptor 2; Interferon gamma receptor accessory factor 1; interferon gamma receptor accessory factor-1; interferon gamma receptor beta chain; Interferon gamma receptor beta-chain; Interferon gamma transducer 1
Gene Aliases: AF-1; IFGR2; IFNGR2; IFNGT1; IMD28
UniProt ID: (Human) P38484
Entrez Gene ID: (Human) 3460
Molecular Function:
transmembrane signal receptor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support