Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Antibody detects endogenous levels of total IFT20.
IFT20 is part of intraflagellar transport (IFT) particles involved in ciliary process assembly. IFT20 may play a role in the trafficking of ciliary membrane proteins from the Golgi complex to the cilium.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: hIFT20; intraflagellar transport 20 homolog; Intraflagellar transport protein 20 homolog; intraflagellar transport protein IFT20; mIFT20; uterine protein
Gene Aliases: 0610009H04Rik; AU015496; IFT20; RGD1309400
UniProt ID: (Human) Q8IY31, (Mouse) Q61025
Entrez Gene ID: (Human) 90410, (Dog) 480620, (Rat) 287541, (Mouse) 55978
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support