Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 3
Antibody detects endogenous levels of total IFT52.
Forms part of a complex involved in intraflagellar transport (IFT), the bi-directional movement of particles required for the assembly, maintenance and functioning of primary cilia.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: intraflagellar transport 52 homolog; Intraflagellar transport protein 52 homolog; Protein NGD5; Protein NGD5 homolog
Gene Aliases: BC037708; C20orf9; CGI-53; IFT52; NGD2; NGD5; RGD1311004
UniProt ID: (Human) Q9Y366, (Mouse) Q62559
Entrez Gene ID: (Human) 51098, (Rat) 362265, (Mouse) 245866
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support