Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
Immunogen sequence: QTYHLDMNET AKQEDILEST TDAAEWSLEV ERVLPQLKVT IRTDNKDWRI HVDQMHQHRS GIESALKETK GFLDKLHNEI TRTLEKISSR EKYINNQLE
HIPPI is required for the formation of cilia. Plays an indirect role in sonic hedgehog signaling, cilia being required for all activity of the hedgehog pathway. Has pro-apoptotic function via its interaction with HIP1, leading to recruit caspase-8 (CASP8) and trigger apoptosis. Has the ability to bind DNA sequence motif 5'-AAAGACATG-3' present in the promoter of caspase genes such as CASP1, CASP8 and CASP10, suggesting that it may act as a transcription regulator; however the relevance of such function remains unclear.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Dermal papilla-derived protein 8; estrogen-related receptor beta like 1; Estrogen-related receptor beta-like protein 1; FLJ10147; HIP1 protein interactor; HIP1-interacting protein; huntingtin interacting protein-1 interacting protein; intraflagellar transport 57 homolog; Intraflagellar transport protein 57 homolog; MHS4R2
Gene Aliases: DERP8; ESRRBL1; HIPPI; IFT57; MHS4R2
UniProt ID: (Human) Q9NWB7
Entrez Gene ID: (Human) 55081
Molecular Function:
structural protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support