Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The proliferation and differentiation of B cells is dependent upon a B-cell antigen receptor (BCR) complex. Binding of antigens to specific B-cell receptors results in a tyrosine phosphorylation reaction through the BCR complex and leads to multiple signal transduction pathways.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: alpha 4; Alpha4 phosphoprotein; B cell signal transduction molecule alpha 4; B-cell signal transduction molecule alpha 4; bA351K23.1 (immunoglobulin binding protein 1 (CD79A) ); CD79a-binding protein 1; immunoglobulin (CD79A) binding protein 1; immunoglobulin binding protein 1; Immunoglobulin-binding protein 1; Lymphocyte signal transduction molecule alpha 4; p52; phosphoprotein, component, 52 kDa; Protein alpha-4; Protein phosphatase 2/4/6 regulatory subunit; protein phosphatase 2A, regulatory subunit alpha-4; Renal carcinoma antigen NY-REN-16
Gene Aliases: AI115586; AI662457; ALPHA-4; AW208785; C81413; IBP1; IGBP1; p52; Pc52; Tap42
UniProt ID: (Human) P78318, (Rat) O08836, (Mouse) Q61249
Entrez Gene ID: (Human) 3476, (Sheep) 101121748, (Rat) 58845, (Mouse) 18518
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support