Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 3
AA Sequence of recombinant protein: AYRPSETLCG GELVDTLQFV CGDRGFYFSR PASRVSRRSR GIVEECCFRS CDLALLETYC ATPAKSE.
Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Human IGF-II. Anti-Human IGF-II-specific antibody was purified by affinity chromatography and then biotinylated.
Sandwich ELISA: To detect Human IGF-II by sandwich ELISA (using 100 µL/well antibody solution) a concentration of 0.25-1.0 µg/mL of this antibody is required. This biotinylated polyclonal antibody, in conjunction with PeproTech Polyclonal Anti-Human IGF-II (500-P12) as a capture antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant Human IGF-II.
Western Blot: To detect hIGF-II by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 µg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant hIGF-II is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions.
500-P12BT-1MG will be provided as 2 x 500 µg
The IGFs are mitogenic, polypeptide growth factors that stimulate the proliferation and survival of various cell types, including muscle, bone, and cartilage tissue in vitro. IGFs are predominantly produced by the liver, although a variety of tissues produce the IGFs at distinctive times. The IGFs belong to the Insulin gene family, which also contains insulin and relaxin. The IGFs are similar to insulin by structure and function, but have a much higher growth-promoting activity than insulin. IGF-II expression is influenced by placenta lactogen, while IGF-I expression is regulated by growth hormone. Both IGF-I and IGF-II signal through the tyrosine kinase type I receptor (IGF-IR), but IGF-II can also signal through the IGF-II/Mannose-6-phosphate receptor. Mature IGFs are generated by proteolytic processing of inactive precursor proteins, which contain N-terminal and C-terminal propeptide regions.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FLJ22066; FLJ44734; H-IGF-2; IGF; IGF-II; Insulin like growth factor; insulin-like growth factor 2 (somatomedin A); Insulin-like growth factor II; insulin-like growth factor type 2; preptin; Somatomedin-A; T3M-11-derived growth factor
Gene Aliases: C11orf43; GRDF; IGF-II; IGF2; PP1446; PP9974
UniProt ID: (Human) P01344
Entrez Gene ID: (Human) 3481
Molecular Function:
growth factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support