Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 4
The recommended shelf life for this product is 1 year from date of receipt.
IGF2BP2 is a member of the IGF-II mRNA-binding protein (IMP) family. It recruits target transcripts to cytoplasmic protein-RNA complexes. The protein contains several four KH domains and two RRM domains. It functions by binding to the 5' UTR of the insulin-like growth factor 2 (IGF2) mRNA and regulating IGF2 translation. Alternate transcriptional splice variants, encoding different isoforms, have been characterized. Autoantibodies against IGF2BP2 are detected in sera from some patients with hepatocellular carcinoma.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Hepatocellular carcinoma autoantigen p62; IGF-II mRNA-binding protein 2; IGF2 mRNA-binding protein 2; insulin-like growth factor 2 mRNA binding protein 2; Insulin-like growth factor 2 mRNA-binding protein 2; insulin-like growth factor 2, binding protein 2; VICKZ family member 2
Gene Aliases: C330012H03Rik; IGF2BP2; IMP-2; IMP2; Neilsen; VICKZ2
UniProt ID: (Human) Q9Y6M1, (Mouse) Q5SF07
Entrez Gene ID: (Human) 10644, (Mouse) 319765
Molecular Function: RNA metabolism protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support