Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
This Antibody was verified by Relative expression to ensure that the antibody binds to the antigen stated.
Immunogen sequence: YKKERRETVI SKLTTCCRRS SNVSYKYSKV NKEEETDENE TEWLMEEIQL PPPRQGKEGQ ENGHITTKSV KALSSLHGDD QDSEDEVLTI PEVKVHSGRG AGAESSHPVR NAQSNALQER EDDRVGLVRG EKARKGKSSS AQQKTVSSTK LVSFHDDSDE DLLHI; Positive Samples: HeLa; Cellular Location: Lysosome membrane, Single-pass type I membrane protein
Lysosomal enzymes containing one or two mannose 6-phosphate (man6P) moieties are moved about in the cell by two distinct but interconnected cycles by means of approximately 300 kDa cation independent mannose 6-phospate receptors (CI-MPR). The biosynthetic cycle refers to the MPR transport of newly synthesized lysosomal enzymes from the trans Golgi network to late endosomes or early lysosomes. The endocytic cycle transports extracellular lysosomal enzymes from the plasma membrane via clathrin-coated vesicles to early endosomes. The entire pool of MPRs cycles between these cellular compartments every 3 hours. The steady state distribution of MPR's is predominantly within late endosomes, fewer in the trans Golgi network and approximately 10 % at cell surface. In addition to its man6P binding activity, the MPR contains a separate binding site for the type II insulin-like growth factor and is capable of binding both man6P and IGF-II simultaneously. An approximately 240 kDa soluble, truncated form, representing the extracellular domain of the protein, has also been found circulating in serum and is capable of binding both ligands.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 300 kDa mannose 6-phosphate receptor; cation-independent mannose 6-phosphate receptor; Cation-independent mannose-6-phosphate receptor; cation-independent MPR; CD222; CI Man-6-P receptor; IGF-II receptor; IGF-II/CI-MPR; Insulin-like growth factor 2 receptor; Insulin-like growth factor II receptor; M6P/IGF2 receptor; M6P/IGF2R; M6PR; mannose-6-phosphate receptor, cation independent; MPR 300
Gene Aliases: AI661837; CD222; CI-M6PR; CI-MPR; CIMPR; IGF2R; Igfr2; M6P-R; M6P/IGF2R; MPR 300; MPR1; MPR300; MPRI
UniProt ID: (Human) P11717, (Mouse) Q07113
Entrez Gene ID: (Human) 3482, (Mouse) 16004, (Rat) 25151
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support